Skip to Content
Merck
All Photos(1)

Key Documents

Y0001657

Quetiapine fumarate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Quetiapine hemifumarate salt, 2-[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethanol hemifumarate, Quetiapine fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N3O2S · 0.5C4H4O4
CAS Number:
Molecular Weight:
441.54
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

quetiapine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\C(O)=O.OCCOCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24.OCCOCCN5CCN(CC5)C6=Nc7ccccc7Sc8ccccc68

InChI

1S/2C21H25N3O2S.C4H4O4/c2*25-14-16-26-15-13-23-9-11-24(12-10-23)21-17-5-1-3-7-19(17)27-20-8-4-2-6-18(20)22-21;5-3(6)1-2-4(7)8/h2*1-8,25H,9-16H2;1-2H,(H,5,6)(H,7,8)/b;;2-1+

InChI key

ZTHJULTYCAQOIJ-WXXKFALUSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Quetiapine fumarate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Quetiapine hemifumarate is a combined serotonin (5HT2) and dopamine (D2) receptor antagonist.
Quetiapine hemifumarate is an atypical antipsychotic, a combined serotonin (5HT2) and dopamine (D2) receptor antagonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Andre Tapp et al.
Drug and alcohol dependence, 149, 18-24 (2015-02-16)
Cocaine addiction continues to be a significant healthcare issue, yet there are no FDA approved medications for the treatment of cocaine use disorder within the United States. This 12-week, prospective, double-blind, randomized, placebo-controlled study examined the effectiveness of quetiapine (Seroquel
Miho Shishikura et al.
Bioscience trends, 8(3), 149-154 (2014-07-18)
It is of importance to determine whether antipsychotic drugs currently prescribed for schizophrenia exert D-amino acid oxidase (DAO)-inhibitory effects. We first investigated whether human (h)DAO can metabolize D-kynurenine (D-KYN) to produce the fluorescent compound kynurenic acid (KYNA) by using high-performance
Piotr Kulinowski et al.
International journal of pharmaceutics, 484(1-2), 235-245 (2015-02-24)
Motivation for the study was the lack of dedicated and effective research and development (R&D) in vitro methods for oral, generic, modified release formulations. The purpose of the research was to assess multimodal in vitro methodology for further bioequivalence study
Susanna Waters et al.
Journal of neural transmission (Vienna, Austria : 1996), 121(11), 1337-1347 (2014-05-13)
The dopaminergic stabilizers pridopidine [4-(3-(methylsulfonyl)phenyl)-1-propylpiperidine] and ordopidine [1-ethyl-4-(2-fluoro-3-(methylsulfonyl)phenyl)piperidine] inhibit psychostimulant-induced hyperactivity, and stimulate behaviour in states of hypoactivity. While both compounds act as dopamine D2 receptor antagonists in vitro, albeit with low affinity, their specific state-dependent behavioural effect profile is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service