Skip to Content
Merck
All Photos(1)

Key Documents

Y0001388

Candesartan cilexetil

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-ethoxy-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-Benzimidazole-7-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, TCV 116, TCY 116

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H34N6O6
CAS Number:
Molecular Weight:
610.66
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

candesartan

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOc1nc2cccc(C(=O)OC(C)OC(=O)OC3CCCCC3)c2n1Cc4ccc(cc4)-c5ccccc5-c6nnn[nH]6

InChI

1S/C33H34N6O6/c1-3-42-32-34-28-15-9-14-27(31(40)43-21(2)44-33(41)45-24-10-5-4-6-11-24)29(28)39(32)20-22-16-18-23(19-17-22)25-12-7-8-13-26(25)30-35-37-38-36-30/h7-9,12-19,21,24H,3-6,10-11,20H2,1-2H3,(H,35,36,37,38)

InChI key

GHOSNRCGJFBJIB-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Candesartan cilexetil EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Candesartan cilexetil is a non-peptide angiotensin (AT2) receptor antagonist.
Candesartan cilexetil is the prodrug form of the potent angiotensin II receptor antagonist, candesartan. The prodrug is cleaved by esterases within the intestine to liberate the active molecule.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2 Oral

Target Organs

Kidney,Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline Fenton et al.
Drugs, 65(4), 537-558 (2005-03-01)
Candesartan cilexetil is the orally administered pro-drug of candesartan, a highly selective antagonist of the angiotensin II subtype 1 receptor that mediates the pressor activities of angiotensin II. Candesartan cilexetil is widely used for the treatment of hypertension and has
T Naka et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 120(12), 1261-1275 (2001-02-24)
Blockade of the action of angiotensin II (AII) has long been a target for the development of novel antihypertensive agents. We recently discovered a novel class of potent nonpeptide AII receptor antagonists, benzimidazole-7-carboxylic acids including candesartan. Candesartan is a highly
Sheridan M Hoy et al.
American journal of cardiovascular drugs : drugs, devices, and other interventions, 10(5), 335-342 (2010-09-24)
Candesartan cilexetil is the orally administered prodrug of candesartan, an angiotensin II subtype 1 receptor antagonist. The pharmacokinetics (area under the plasma concentration-time curve and maximum plasma concentration) of candesartan do not appear to be affected by age, sex, or
J M Mallion et al.
Journal of human hypertension, 14 Suppl 2, S33-S41 (2000-11-22)
Candesartan is one of the first angiotensin II receptor antagonists (AIIRAs) to be developed. It binds tightly to and dissociates slowly from the angiotensin subtype 1 (AT1) receptor in vitro. These binding characteristics differ from those of losartan, which demonstrates
Stephanie E Easthope et al.
Drugs, 62(8), 1253-1287 (2002-05-16)
Candesartan cilexetil is converted to the angiotensin II receptor antagonist candesartan during absorption from the gastrointestinal tract. The selective and competitive binding of candesartan to the angiotensin II type 1 (AT(1)) receptor prevents binding of angiotensin II, a key mediator

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service