Skip to Content
Merck
All Photos(1)

Key Documents

Y0001256

Levetiracetam impurity C

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Hydroxypyridine, 2(1H)-Pyridone, 2-Pyridinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO
CAS Number:
Molecular Weight:
95.10
Beilstein:
105757
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levetiracetam

manufacturer/tradename

EDQM

bp

280-281 °C (lit.)

mp

105-107 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1ccccn1

InChI

1S/C5H5NO/c7-5-3-1-2-4-6-5/h1-4H,(H,6,7)

InChI key

UBQKCCHYAOITMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levetiracetam impurity C EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Ken-ichi Kusakabe et al.
Bioorganic & medicinal chemistry, 21(7), 2045-2055 (2013-02-12)
Selective CB2 agonists have the potential for treating pain without central CB1-mediated adverse effects. Screening efforts identified 1,2-dihydro-3-isoquinolone 1; however, this compound has the drawbacks of being difficult to synthesize with two asymmetric carbons on an isoquinolone scaffold and of
Christian Wiegand et al.
Chemical communications (Cambridge, England), 48(82), 10195-10197 (2012-09-11)
3-Substituted 2-pyridones were enantioselectively (68-90% ee) converted into the respective 3-hydroxypyridine-2,6-diones by a sequence consisting of a template-mediated type II photooxygenation and an acid-catalysed rearrangement.
Mingwen Zhu et al.
Chemical communications (Cambridge, England), 47(48), 12876-12878 (2011-11-15)
A highly efficient and convenient CuCl/2-pyridonate catalytic system for oxidative homocoupling of azoles affording a biazole product has been developed. With this system, a variety of biazoles have been effectively synthesized in good to excellent yields in the presence of
Mettu Ravinder et al.
Bioorganic & medicinal chemistry letters, 22(18), 6010-6015 (2012-08-18)
Twenty-six 2-pyridone derivatives (8a-8z), which are structurally analogous to amrinone and milrinone two important cardiotonic drugs, are synthesized and characterized. The synthesis of 2-pyridone derivatives involves addition, followed by cyclization between Baylis-Hillman acetates (7a-7k) and enamino esters or nitriles (3a-3e).
Susan Blaser et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 12(10), 1841-1850 (2011-05-25)
A combined spectroscopic and ab initio theoretical study of the doubly hydrogen-bonded complex of 2-pyridone (2PY) with NH(3) has been performed. The S(1)←S(0) spectrum extends up to ≈1200 cm(-1) above the 0(0) (0) band, close to twice the range observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service