Skip to Content
Merck
All Photos(1)

Key Documents

Y0001209

Meglumine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-Methyl-D-glucamine, 1-Deoxy-1-(methylamino)-D-glucitol, Meglumine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO5
CAS Number:
Molecular Weight:
195.21
Beilstein:
385906
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

meglumine

autoignition temp.

~662 °F

manufacturer/tradename

EDQM

mp

129-131.5 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C7H17NO5/c1-8-2-4(10)6(12)7(13)5(11)3-9/h4-13H,2-3H2,1H3/t4-,5+,6+,7+/m0/s1

InChI key

MBBZMMPHUWSWHV-BDVNFPICSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Meglumine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrik Zamecnik et al.
Radiology, 273(3), 879-886 (2014-07-26)
To assess the feasibility of automatic needle-guide tracking by using a real-time phase-only cross correlation ( POCC phase-only cross correlation ) algorithm-based sequence for transrectal 3-T in-bore magnetic resonance (MR)-guided prostate biopsies. This study was approved by the ethics review
Chong Hyun Suh et al.
Radiology, 272(2), 504-513 (2014-04-05)
To determine the utility of intravoxel incoherent motion (IVIM)-derived perfusion and diffusion parameters for differentiation of atypical primary central nervous system lymphoma (PCNSL) from glioblastoma in patients who do not have acquired immunodeficiency syndrome. The institutional review board approved this
Philipp Kickingereder et al.
Radiology, 272(3), 843-850 (2014-05-13)
To compare multiparametric diagnostic performance with diffusion-weighted, dynamic susceptibility-weighted contrast material-enhanced perfusion-weighted, and susceptibility-weighted magnetic resonance (MR) imaging for differentiating primary central nervous system lymphoma (PCNSL) and atypical glioblastoma. This retrospective study was institutional review board-approved and informed consent was
Gilles Barone-Rochette et al.
Journal of magnetic resonance imaging : JMRI, 40(6), 1347-1354 (2013-12-03)
To determine the value of cardiac MRI for the monitoring and the prognosis of patients with acute myocarditis. Cardiac MRI plays an increasingly important role in the diagnosis of acute myocarditis. However, it is less established as a prognostic tool
Lourens F H J Robbers et al.
Radiology, 272(1), 113-122 (2014-03-13)
To investigate the effects of cell therapy on myocardial perfusion recovery after treatment of acute myocardial infarction (MI) with primary percutaneous coronary intervention (PCI). In this HEBE trial substudy, which was approved by the institutional review board (trial registry number

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service