Skip to Content
Merck
All Photos(1)

Documents

Y0001203

Hesperidin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3′,5,7-Trihydroxy 4′-methoxyflavanone 7-rutinoside, Hesperetin 7-rhamnoglucoside, Hesperitin-7-rutinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H34O15
CAS Number:
Molecular Weight:
610.56
Beilstein:
75140
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

hesperidin

manufacturer/tradename

EDQM

mp

250-255 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1O)[C@@H]2CC(=O)c3c(O)cc(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2

InChI

1S/C28H34O15/c1-10-21(32)23(34)25(36)27(40-10)39-9-19-22(33)24(35)26(37)28(43-19)41-12-6-14(30)20-15(31)8-17(42-18(20)7-12)11-3-4-16(38-2)13(29)5-11/h3-7,10,17,19,21-30,32-37H,8-9H2,1-2H3/t10-,17-,19+,21-,22+,23+,24-,25+,26+,27+,28+/m0/s1

InChI key

QUQPHWDTPGMPEX-QJBIFVCTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Studies of the behavior of hesperidin and hesperetin (the aglycone) with DMPC liposomes find the aglycone penetrates more deeply and binds more strongly to the "membrane." This may explain the better bioavailability of bioflavonoid aglycones in general.

Biochem/physiol Actions

Flavonoid phytochemical extracted from citrus species. It is the rhamnoglucoside (rutinoside) of hesperetin.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuji Tanaka et al.
Journal of biomedicine & biotechnology, 2012, 516981-516981 (2011-12-17)
β-Cryptoxanthin, a carotenoid, and hesperidin, a flavonoid, possess inhibitory effects on carcinogenesis in several tissues. We recently have prepared a pulp (CHRP) and citrus juices (MJ2 and MJ5) from a satsuma mandarin (Citrus unshiu Mar.) juice (MJ). They contain high
A Garg et al.
Phytotherapy research : PTR, 15(8), 655-669 (2001-12-18)
Hesperidin, a bioflavonoid, is an abundant and inexpensive by-product of Citrus cultivation. A deficiency of this substance in the diet has been linked with abnormal capillary leakiness as well as pain in the extremities causing aches, weakness and night leg
Asjad Visnagri et al.
Pharmaceutical biology, 52(7), 814-828 (2014-02-25)
Diabetic neuropathy (DN) is one of the most common long-term complications of diabetes mellitus and clinically can be characterized by an elevated nociceptive response with electrophysiological conduction abnormalities. The present investigation was designed to evaluate the neuroprotective effect of hesperidin
Silvia Yumnam et al.
PloS one, 9(6), e101321-e101321 (2014-07-01)
Hesperidin, a natural flavonoid abundantly present in Citrus is known for its anti-cancer, anti-oxidant and anti-inflammatory properties. In this study we examined the effect of hesperidin on HepG2 cells. HepG2 cells treated with various concentration of hesperidin undergo a distinct
G Allam et al.
Journal of helminthology, 88(3), 362-370 (2013-05-10)
Hesperidin has been reported to exert a wide range of pharmacological effects, including antifungal, antiviral, antioxidant, anti-inflammatory and anticarcinogenic activities. Herein, the schistosomicidal activity of this compound was evaluated in vitro and in vivo. Using an in vitro assay, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service