Skip to Content
Merck
All Photos(1)

Key Documents

Y0001194

Resveratrol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3,4′,5-Trihydroxy-trans-stilbene, 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

resveratrol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)\C=C\c2cc(O)cc(O)c2

InChI

1S/C14H12O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h1-9,15-17H/b2-1+

InChI key

LUKBXSAWLPMMSZ-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Selective inhibitor of COX−1.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>392.0 °F - closed cup - (External MSDS)

Flash Point(C)

> 200.0 °C - closed cup - (External MSDS)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agneta Oskarsson et al.
The Prostate, 74(8), 839-851 (2014-03-13)
Resveratrol (RSV) and resveratrol analogs have a potential use in prostate cancer chemoprevention due to effects on for example, cell growth, apoptosis, angiogenesis, and metastasis. However, inhibition of CYP17A1, a key enzyme in the androgen biosynthesis and a target for
Dominique Delmas et al.
Annals of the New York Academy of Sciences, 1290, 90-97 (2013-07-17)
The preventive effects of the phytoalexin trans-resveratrol toward cancer have been largely described at the cellular and molecular levels in both in vivo and in vitro models; however, its primary targets are still poorly identified. In this review, we show
Morten Møller Poulsen et al.
Annals of the New York Academy of Sciences, 1290, 74-82 (2013-07-17)
In the search for novel preventive and therapeutic modalities in the management of metabolic diseases and obesity, resveratrol has attracted great attention over the past decades. Preclinical trials suggest that resveratrol mimics the metabolic effects of calorie restriction (CR) via
Silvie Timmers et al.
Annals of the New York Academy of Sciences, 1290, 83-89 (2013-07-17)
The number of people suffering from metabolic disorders is dramatically increasing worldwide. The need for new therapeutic strategies to combat this growing epidemic of metabolic diseases is therefore also increasing. In 2003, resveratrol was discovered to be a small molecule
Mary Ann Augustin et al.
Annals of the New York Academy of Sciences, 1290, 107-112 (2013-07-17)
There has been interest in the use of trans-resveratrol as a natural preventative agent for improving health and alleviating a range of diseases. However, resveratrol has low bioavailability, and this has been associated with its poor water solubility, its low

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service