Skip to Content
Merck
All Photos(1)

Key Documents

Y0001105

Drospirenone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6β,7β:15β,16β -Dimethylene-3-oxo-17α-pregn-4-ene-21,17-carbolactone, Dihydrospirorenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30O3
CAS Number:
Molecular Weight:
366.49
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

drospirenone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@]12CCC(=O)C=C1[C@@H]3C[C@@H]3[C@@H]4[C@@H]2CC[C@@]5(C)[C@H]4[C@@H]6C[C@@H]6[C@@]57CCC(=O)O7

InChI

1S/C24H30O3/c1-22-6-3-12(25)9-17(22)13-10-14(13)20-16(22)4-7-23(2)21(20)15-11-18(15)24(23)8-5-19(26)27-24/h9,13-16,18,20-21H,3-8,10-11H2,1-2H3/t13-,14+,15-,16+,18+,20-,21+,22-,23+,24+/m1/s1

InChI key

METQSPRSQINEEU-HXCATZOESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Drospirenone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Drospirenone is a fourth-generation progestin that has antimineralocorticoid, and antiandrogenic activity in addition to potent progestogenic activity. In two recent studies drospirenone appeared to double the risk of venous thromboembolism compared to levonorgestrel, although other studies found little added risk.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L M Lopez et al.
The Cochrane database of systematic reviews, (1)(1), CD006586-CD006586 (2008-02-07)
Premenstrual syndrome (PMS) is a common problem. Premenstrual dysphoric disorder (PMDD) is a severe form of PMS. Combined oral contraceptives (COCs), which have both progestin and estrogen, have been examined for their ability to relieve premenstrual symptoms. A COC containing
T Simoncini et al.
Climacteric : the journal of the International Menopause Society, 13(1), 22-33 (2009-11-27)
Hormone therapy is effective for the relief of menopausal symptoms. For women with a uterus, addition of a progestogen is required to protect the endometrium. However, synthetic progestins differ in certain pharmacological characteristics that may have implications for clinical practice.
Cassiana R G Giribela et al.
Contraception, 86(1), 35-41 (2012-04-03)
Combined oral contraceptives (COCs) may lead to a rise in cardiovascular disease risk, possibly associated with changes in blood pressure and endothelial function. The objective was to evaluate the impact of COC containing 20 mcg of ethinylestradiol (EE) and 3
Naida Sehovic et al.
The Annals of pharmacotherapy, 44(5), 898-903 (2010-04-08)
To review primary literature regarding the risk of venous thromboembolism (VTE) in users of combined oral contraceptives (COCs) containing drospirenone compared to COCs containing other progestins. A literature search of MEDLINE and EMBASE (1950-January 2010) was conducted using the following
A R Genazzani et al.
Climacteric : the journal of the International Menopause Society, 10 Suppl 1, 11-18 (2007-03-17)
Drospirenone is a unique progestogen derived from 17alpha-spirolactone, with a pharmacologic profile very similar to that of endogenous progesterone. In contrast with other available progestins, drospirenone is a progestogen with aldosterone receptor antagonism (PARA) through its affinity for the mineralocorticoid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service