Skip to Content
Merck
All Photos(1)

Key Documents

Y0000802

Dipyridamole for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dipyridamole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40N8O4
CAS Number:
Molecular Weight:
504.63
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dipyridamole

manufacturer/tradename

EDQM

mp

165-166 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OCCN(CCO)c1nc(N2CCCCC2)c3nc(nc(N4CCCCC4)c3n1)N(CCO)CCO

InChI

1S/C24H40N8O4/c33-15-11-31(12-16-34)23-26-20-19(21(27-23)29-7-3-1-4-8-29)25-24(32(13-17-35)14-18-36)28-22(20)30-9-5-2-6-10-30/h33-36H,1-18H2

InChI key

IZEKFCXSFNUWAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dipyridamole for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Selective inhibitor of phosphodiesterase V (PDE 5); potent coronary vasodilator drug; adenosine transport inhibitor; inhibitor of platelet aggregation.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wolfgang G Eisert
Advances in cardiology, 47, 78-86 (2012-08-22)
The antithrombotic activity of dipyridamole was initially discovered in an in vivo experiment about half a century ago. At that time science had not appreciated the complexity of the regulation of local thrombus formation. Inhibition of platelets has been the
E L L M De Schryver et al.
The Cochrane database of systematic reviews, (3)(3), CD001820-CD001820 (2007-07-20)
Patients with limited cerebral ischaemia of arterial origin are at risk of serious vascular events (4% to 11% annually). Aspirin reduces that risk by 13%. In one trial, adding dipyridamole to aspirin was associated with a 22% risk reduction compared
Subrata Chakrabarti et al.
Vascular pharmacology, 48(4-6), 143-149 (2008-03-18)
Stroke is a leading cause of morbidity and mortality and the most common cause of neurological disability in older individuals. Prevention of recurrent stroke includes risk factor modification as well as the use of therapies that inhibit platelet activation. One
J Greenhalgh et al.
Health technology assessment (Winchester, England), 15(31), 1-178 (2011-09-06)
Occlusive vascular events such as myocardial infarction (MI), ischaemic stroke and transient ischaemic attack (TIA) are the result of a reduction in blood flow associated with an artery becoming narrow or blocked through atherosclerosis and atherothrombosis. Peripheral arterial disease is
Hyung-Hwan Kim et al.
Arteriosclerosis, thrombosis, and vascular biology, 28(3), s39-s42 (2008-01-05)
Dipyridamole (DP) is a phosphodiesterase inhibitor that increases the intracellular levels of cyclic adenosine monophosphate (cAMP) and cyclic guanine monophosphate (cGMP) by preventing their conversion to AMP and GMP, respectively. By increasing cAMP and cGMP levels in platelets, DP reversibly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service