Skip to Content
Merck
All Photos(1)

Key Documents

Y0000792

Poly(vinyl acetate) dispersion 30 per cent

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Poly(vinyl acetate) dispersion

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C4H6O2]n
CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

polyvinyl acetate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O(C(=O)C)C=C

InChI

1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3

InChI key

XTXRWKRVRITETP-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Poly(vinyl acetate) dispersion 30 per cent EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Congming Xiao et al.
International journal of biological macromolecules, 52, 349-352 (2012-10-31)
Tailor-made conjunct of methyl cellulose (MC) and polyvinyl acetate (PVAc) was synthesized through the combination of reversible addition-fragmentation chain transfer (RAFT) polymerization and thiol-ene click reaction. MC was firstly transferred into unsaturated MC (UMC), and then covalently connected with well-defined
Il Gyu Kang et al.
American journal of rhinology & allergy, 24(5), 392-395 (2011-01-20)
The aim of this study was to evaluate the effect of repeated expandable polyvinyl acetate (EPA) packing for preventing stenosis of the frontal sinus ostium after patients undergo functional endoscopic sinus surgery (FESS). A nonrandomized, prospective study of 20 patients
V Andonova et al.
Die Pharmazie, 67(7), 601-604 (2012-08-15)
During the last decade the number of investigations on the preparation and application of more effective drug release systems on the basis of nanocarriers from biocompatible and biodegradable polymers are considerably increasing. This is notably in force for practically water
Seon Tae Kim et al.
International journal of pediatric otorhinolaryngology, 77(1), 113-116 (2012-11-08)
The purpose of this prospective study was to determine the effectiveness of polyurethane foam (PUF) and polyvinyl acetate (PA) as packing materials for reducing post-conchotomy bleeding, pain, and headaches. This study was a prospective, randomized and single-blinded controlled study. Fifty-two
Morphology of new zwitterionic copolymer matrices for sustained drug release.
Bistra Kostova et al.
Journal of controlled release : official journal of the Controlled Release Society, 148(1), e44-e45 (2011-05-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service