Skip to Content
Merck
All Photos(1)

Key Documents

Y0000687

Alverine citrate salt

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-Ethyl-3,3′-diphenyldipropylamine, NSC 35459

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27N · C6H8O7
CAS Number:
Molecular Weight:
473.56
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

alverine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CCCc1ccccc1)CCCc2ccccc2

InChI

1S/C20H27N.C6H8O7/c1-2-21(17-9-15-19-11-5-3-6-12-19)18-10-16-20-13-7-4-8-14-20;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-8,11-14H,2,9-10,15-18H2,1H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

RYHCACJBKCOBTJ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Used for studies of pharmacological activity including:
  • Synergism with fluconazole against Saccharomyces cerevisiae and Candida albicans
  • Efficacy of combination with simeticone on abdominal pain associated with irritable bowel syndrome
  • Nutrient-sensitized screening for drugs that shift energy metabolism from mitochondrial respiration to glycolysis
  • Drug effects viewed from a signal transduction network perspective
  • Enhanced cytotoxic effect of proteasome inhibitor MG132
  • Mechanism of action on phasic smooth muscles

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chinmoy Ghosh et al.
Drug testing and analysis, 2(6), 284-291 (2010-06-22)
A rapid and highly sensitive method for the determination of alverine (ALV) and its metabolite, para hydroxy alverine (PHA), in human plasma using LC-MS/MS in positive ion electrospray ionization (ESI) in multiple reactions monitoring (MRM) mode was developed and validated.
Engin Altintaş et al.
The Turkish journal of gastroenterology : the official journal of Turkish Society of Gastroenterology, 19(3), 174-179 (2008-12-31)
Successful colonoscopy depends on the insertion of the instrument to the cecum, a detailed examination, and minimal discomfort to the patient during the procedure. The aim of this study was to determine the effects of alverine citrate plus simethicone on
Samia Ammar et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 16(12), 1156-1161 (2009-05-01)
A sesquiterpenoid Bakkenolide (1), and two steroids, (3beta, 22E)-Stigmasta-5, 22-diène-3-ol (Stigmasterol) (2) and stigmasterol 3beta-glucoside (3), isolated from the Hertia cheirifolia (L.) chloroform extract, were evaluated respectively for their spasmolytic and anti-inflammatory activities. We note that these natural products were
A Abysique et al.
Alimentary pharmacology & therapeutics, 13(4), 561-566 (1999-04-24)
Alverine citrate is commonly used in the treatment of painful affections of the colon. To determine whether alverine citrate acts on the vagal sensory endings. Unitary recordings were performed at the level of the vagal fibres in the nodose ganglion
A general practice study to compare alverine citrate with mebeverine hydrochloride in the treatment of irritable bowel syndrome.
G J Tudor
The British journal of clinical practice, 40(7), 276-278 (1986-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service