Skip to Content
Merck
All Photos(1)

Key Documents

Y0000550

Sulbactam for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sulbactam, (2S,5R)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide, Betamaze, CP 45899, CP-45,899, Penicillanic acid 1,1-dioxide, Penicillanic acid S,S-dioxide, Penicillanic acid dioxide, Penicillanic acid sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO5S
CAS Number:
Molecular Weight:
233.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulbactam

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O

InChI

1S/C8H11NO5S/c1-8(2)6(7(11)12)9-4(10)3-5(9)15(8,13)14/h5-6H,3H2,1-2H3,(H,11,12)/t5-,6+/m1/s1

InChI key

FKENQMMABCRJMK-RITPCOANSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulbactam for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu Xia et al.
PloS one, 9(7), e100707-e100707 (2014-07-02)
To investigate CTX-M genotypes among extended-spectrum β-lactamase-producing Escherichia coli (ESBL-EC) isolated from patients with community-onset and hospital-onset infections in China, their clonality and the distribution of CTX-M variants in different specimens of community-onset and hospital-onset infections. ESBL-EC isolates were collected
A S Levin
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 8(3), 144-153 (2002-05-16)
Recent studies have highlighted the emergence of infections involving multiresistant Acinetobacter clinical isolates. Sulbactam offers direct antimicrobial activity against Acinetobacter species. Accordingly, co-administration of sulbactam with ampicillin or cefoperazone offers the potential of effective empirical therapy against Acinetobacter and other
G S Singh
Mini reviews in medicinal chemistry, 4(1), 93-109 (2004-02-03)
Beta-lactam ring-containing compounds such as penicillins, ampicillin, amoxicillin, cephalosporins and carbapenems are among the most famous antibiotics. This article reviews the recent developments in study of cephems, oxacephems, penams and sulbactam. Many of the compounds reviewed have potential antibacterial activity
J D Williams
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 24(3), 494-497 (1997-03-01)
beta-Lactamase inhibitors such as sulbactam are beta-lactam compounds that have low antimicrobial activity but are able to inhibit enzymes (beta-lactamases) that destroy beta-lactam antibiotics like penicillins and cephalosporins. The main activity of beta-lactamase inhibitors is directed against plasmid-mediated transferable enzymes
M Akova
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 14 Suppl 1, 185-188 (2007-12-25)
Sulbactam irreversibly inhibits the hydrolytic activity of beta-lactamases. This compound is commercially available in combination with either ampicillin or cefoperazone. In each instance, the activity of the partner antibiotic against beta-lactamase-producing bacteria is restored. One of the particular advantages of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service