Skip to Content
Merck
All Photos(1)

Key Documents

Y0000486

Ketorolac trometamol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ketorolac tris salt, (±)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid tris salt, Toradol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO3 · C4H11NO3
CAS Number:
Molecular Weight:
376.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ketorolac

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(CO)(CO)CO.OC(=O)C1CCn2c1ccc2C(=O)c3ccccc3

InChI

1S/C15H13NO3.C4H11NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13;5-4(1-6,2-7)3-8/h1-7,11H,8-9H2,(H,18,19);6-8H,1-3,5H2

InChI key

BWHLPLXXIDYSNW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ketorolac trometamol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Pfaffenrath et al.
Cephalalgia : an international journal of headache, 32(10), 766-777 (2012-06-20)
Ketorolac is a non-triptan, non-opioid, mixed cyclooxygenase (COX)1/2-inhibitor for short-term management of moderate-to-severe acute pain. This trial evaluated an intranasal formulation of ketorolac tromethamine (SPRIX®) containing 6% lidocaine (ROX-828) for the acute treatment of migraine with and without aura as
Stephen J Kim et al.
Archives of ophthalmology (Chicago, Ill. : 1960), 130(4), 456-460 (2012-04-12)
To investigate the adverse ocular effects of intravitreal ketorolac (4 mg) in patients with chronic uveitis and complications of chronic inflammation (macular edema). We conducted a prospective phase 1 clinical trial involving 10 eyes of 10 adult patients with chronic
Giuseppina Magni et al.
Anesthesia and analgesia, 116(2), 443-447 (2013-01-11)
Ketorolac tromethamine (ketorolac) is a nonsedating drug with potent analgesic and moderate anti-inflammatory activity, which does not increase the sedation level. The safety of ketorolac with respect to risk of bleeding has been demonstrated in large numbers of patients undergoing
Michel Weber et al.
Acta ophthalmologica, 91(1), e15-e21 (2012-09-14)
To determine whether indomethacin 0.1% eye drops are at least as effective as ketorolac 0.5% eye drops in treating ocular inflammation following cataract surgery. Prospective, multicenter, investigator-masked, parallel-group, randomized, active-controlled clinical trial. Cataract patients were randomized in a 1:1 ratio
Meizi Wang et al.
Retina (Philadelphia, Pa.), 32(10), 2158-2164 (2012-10-27)
To investigate the concentrations and pharmacokinetics of ketorolac in the rabbits by three different routes of administrations: a single intracameral, intravitreal, and suprachoroidal injection. Fifty-four New Zealand white rabbits received ketorolac (250 μg/0.05 mL) in one eye by a single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service