Skip to Content
Merck
All Photos(1)

Key Documents

Y0000471

Buspirone for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Buspirone hydrochloride, N-[4-[4-(2-Pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5]decane-7,9-dione hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31N5O2 · HCl
CAS Number:
Molecular Weight:
421.96
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

buspirone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].O=C1CC2(CCCC2)CC(=O)N1CCCCN3CCN(CC3)c4ncccn4

InChI

1S/C21H31N5O2.ClH/c27-18-16-21(6-1-2-7-21)17-19(28)26(18)11-4-3-10-24-12-14-25(15-13-24)20-22-8-5-9-23-20;/h5,8-9H,1-4,6-7,10-17H2;1H

InChI key

RICLFGYGYQXUFH-UHFFFAOYSA-N

Gene Information

human ... HTR1A(3350)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Buspirone for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

5-HT1A serotonin receptor agonist; anxiolytic.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kah-Chung Leong et al.
Behavioural brain research, 234(2), 299-302 (2012-07-18)
Previous findings indicate that post-training administration of the anxiogenic α(2)-adrenoceptor antagonist RS 79948-197 facilitates the consolidation of dorsal striatal-dependent habit memory. The present study examined the effect of concurrent administration of the anxiolytic serotonin 5-HT(1A) receptor partial agonist buspirone on
Long Yuan et al.
Bioanalysis, 4(23), 2795-2804 (2012-12-12)
A dried blood spot (DBS) bioanalysis assay involves many steps, such as the preparation of standard (STD) and QC samples in blood, the spotting onto DBS cards, and the cutting-out of the spots. These steps are labor intensive and time
Jack Bergman et al.
The international journal of neuropsychopharmacology, 16(2), 445-458 (2012-07-26)
Converging lines of evidence indicate that elevations in synaptic dopamine levels play a pivotal role in the reinforcing effects of cocaine, which are associated with its abuse liability. This evidence has led to the exploration of dopamine receptor blockers as
Feng Qiu et al.
Biomedical chromatography : BMC, 27(2), 233-239 (2012-06-19)
This study firstly describes the development of an accurate and sensitive high-performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) assay for the quantification of Taiwanin E methyl ether (TEME) in rat plasma. The assay involved a simple liquid-liquid extraction step with ethyl
Repurposing buspirone for drug addiction treatment.
Bernard Le Foll et al.
The international journal of neuropsychopharmacology, 16(2), 251-253 (2012-11-24)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service