Skip to Content
Merck
All Photos(1)

Key Documents

Y0000443

Flunixin meglumine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-[[2-Methyl-3-(trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic acid meglumine salt, Banamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11F3N2O2·C7H17NO5
CAS Number:
Molecular Weight:
491.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

flunixin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.Cc1c(Nc2ncccc2C(O)=O)cccc1C(F)(F)F

InChI

1S/C14H11F3N2O2.C7H17NO5/c1-8-10(14(15,16)17)5-2-6-11(8)19-12-9(13(20)21)4-3-7-18-12;1-8-2-4(10)6(12)7(13)5(11)3-9/h2-7H,1H3,(H,18,19)(H,20,21);4-13H,2-3H2,1H3/t;4-,5+,6+,7+/m.0/s1

InChI key

MGCCHNLNRBULBU-WZTVWXICSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Flunixin meglumine (IC50 = 1 nM) can be used as a drug for animals for the management of intestinal ischaemia, colic, and endotoxemia in horses.

Biochem/physiol Actions

COX1, COX2 and PLA2 inhibitor non-narcotic analgesic, anti-pyretic, anti-inflammatory.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Morton et al.
Equine veterinary journal. Supplement, (39)(39), 112-116 (2011-08-04)
The effects of prostaglandins and nonsteroidal anti-inflammatory drugs (NSAIDs) on repair of equine intestinal mucosa are important since most horses with gastrointestinal diseases are routinely treated with NSAIDs, such as flunixin meglumine (FM), and these drugs can be toxic to
Pharmacokinetic interactions of flunixin meglumine and doxycycline in broiler chickens.
F Yang et al.
Journal of veterinary pharmacology and therapeutics, 36(1), 85-88 (2012-02-02)
J F Marshall et al.
Equine veterinary journal. Supplement, (39)(39), 106-111 (2011-08-04)
Flunixin meglumine is used for treatment of equine colic despite evidence of inhibited recovery of mucosal barrier function following small intestinal ischaemic injury. This study aimed to characterise an alternative treatment (AHI-805) for abdominal pain in the horse. To determine
J Cuervo-Arango et al.
Theriogenology, 77(3), 514-524 (2011-10-01)
The development of hemorrhagic anovulatory follicles (HAF) involves luteinization and hemorrhage of the follicle. This is observed on ultrasound as an increase in the echogenicity of the granulosa layer and formation of echoic particles in the antrum. The inhibition of
Mariana S C F Silva et al.
Behavioural brain research, 271, 249-257 (2014-06-18)
Corticotropin-releasing factor (CRF) plays a critical role in the mediation of physiological and behavioral responses to stressors. In the present study, we investigated the role played by the CRF system within the dorsomedial hypothalamus (DMH) in the modulation of anxiety-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service