Skip to Content
Merck
All Photos(3)

Key Documents

W374400

Sigma-Aldrich

Massoia Lactone

greener alternative

natural, ≥95%, FG

Synonym(s):

6-Pentyl-5,6-dihydropyran-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O2
CAS Number:
Molecular Weight:
168.23
FEMA Number:
3744
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.037
NACRES:
NA.21
Organoleptic:
coconut; creamy; fatty; nutty; fruity
grade:
FG
Halal
Kosher
natural
biological source:
massoia bark (oil)
food allergen:
no known allergens

biological source

massoia bark (oil)

Quality Level

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥95%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

coconut; creamy; fatty; nutty; fruity

SMILES string

O=C1C=CCC(CCCCC)O1

InChI

1S/C10H16O2/c1-2-3-4-6-9-7-5-8-10(11)12-9/h5,8-9H,2-4,6-7H2,1H3

InChI key

NEDIAPMWNCQWNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service