UPH2O
Ultra pure DNA-free Water
DNA-free, Nuclease-free water
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106229
Recommended Products
Quality Level
form
liquid
feature
DNA free
RNase and DNase free (also Nickase free)
packaging
pkg of 1.5 mL
storage condition
room temperature
technique(s)
DNA sequencing: suitable
RT-PCR: suitable
cDNA synthesis: suitable
qPCR: suitable
dilution
(Molecular assays)
shipped in
ambient
General description
Ultra pure DNA-free Water is generated using a Milli-Q® IQ 7000 Ultrapure Water Purification System, filter-sterilized and UV-treated, and undergoes stringent quality control testing to ensure that it is free of detectable levels of DNA, DNase, RNase, and Nickase to reduce the risk of false positive and false negative results in molecular diagnostic assays. Ultra pure DNA-free Water should be used with other ultra pure DNA-free reagents, such as Ultra pure JumpStart™ Taq DNA Polymerase, Ultra pure High-temp Reverse Transcriptase, and accompanying Ultra pure RT-PCR buffers, to minimize false positive and negative results.
Ultra pure DNA-free Water was designed for use in highly-regulated industries in applications that require a high level of performance, lot-to-lot consistency, and high sensitivity without false results from reagent-borne contamination while meeting industry-driven critical regulations. To meet this requirements we have increased the quality level of our products. The M-Clarity Program defines product quality. It provides transparency so that you can choose, with confidence, suitable products that meet your needs with respect to quality and regulatory compliance, transparency, change notification, and documentation.
Ultra pure DNA-free Water was designed for use in highly-regulated industries in applications that require a high level of performance, lot-to-lot consistency, and high sensitivity without false results from reagent-borne contamination while meeting industry-driven critical regulations. To meet this requirements we have increased the quality level of our products. The M-Clarity Program defines product quality. It provides transparency so that you can choose, with confidence, suitable products that meet your needs with respect to quality and regulatory compliance, transparency, change notification, and documentation.
Application
Ultra pure DNA-free Water can be used for:
- Use in molecular assays
- DNA sequencing
- cDNA synthesis
- PCR and qPCR
- For further manufacture
Features and Benefits
- DNA-free
- DNase-free, RNase-free, and nickase-free
- Filter-sterilized
- UV-treated, not DEPC-treated
- Ideal for high throughput molecular applications
- Enhanced change control and quality agreement
Packaging
Ultra pure DNA-free Water is provided in larger SKU sizes for ease of use in high throughput RT-PCR applications and for further manufacture. Ultra pure DNA-free Water should be used with other ultra pure DNA-free reagents, such as Ultra pure JumpStart™ Taq DNA Polymerase, Ultra pure High-temp Reverse Transcriptase, and accompanying Ultra pure RT-PCR buffers, to minimize false positive and negative results.
Other Notes
Quality testing ensures the absence of detectable levels of contaminating DNA (<=1 copy of prokaryotic gDNA, <=0.1 copy of eukaryotic gDNA, and <=10 copies of plasmid DNA per 100 units of enzyme) as detected by qPCR using universal primers against 16S capable of detecting > 500,000 prokayotes, 18S capable of detecting > 110,000 eukaryotes, and Ori capable of detecting > 25,000 plasmids, respectively.
Legal Information
JumpStart is a trademark of Sigma-Aldrich Co. LLC
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
nwg
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service