Skip to Content
Merck
All Photos(3)

Key Documents

T8533

Sigma-Aldrich

o-Tolidine

≥95% purity, powder or chunks

Synonym(s):

3,3′-Dimethylbenzidine, 4,4′-Bianisidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-C6H3(CH3)-4-NH2]2
CAS Number:
Molecular Weight:
212.29
Beilstein:
2210640
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

o-Tolidine, ≥95%

Quality Level

Assay

≥95%

form

powder or chunks

technique(s)

titration: suitable

color

white to light brown

mp

128-131 °C (lit.)

solubility

water: slightly soluble 1.3 g/L at 25 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1cc(ccc1N)-c2ccc(N)c(C)c2

InChI

1S/C14H16N2/c1-9-7-11(3-5-13(9)15)12-4-6-14(16)10(2)8-12/h3-8H,15-16H2,1-2H3

InChI key

NUIURNJTPRWVAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Tolidine is a diacidic dye which is sparingly soluble in water and easily soluble in ether and alcohol. On exposure to chlorine or bromine vapors, it forms an intense blue color. This forms the basis for detection of chlorine, nitrates and hydrogen peroxide in air or water. It turns yellow on reacting with free chlorine. o-Tolidine easily forms acid salts.

Application

o-Tolidine has also been used to determine the presence of laccase activity in T. versicolor fungal colonies.
Sensitive colorimetric reagent for gold and for free chlorine in water.

Warning

May be carcinogenic.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification and Characterization of Laccase from the
White Rot Fungus Trametes versicolor
Moon-Jeong
Journal of Microbiology (2005)
Wiley-VCH
Ullmann's Fine Chemicals null
Leo M.L. Nollet
Handbook of Water Analysis (2013)
3,3'-Dimethylbenzidine.
Report on carcinogens : carcinogen profiles, 10, 104-105 (2004-08-25)
Dyes metabolized to 3,3'-dimethylbenzidine (3,3'-dimethylbenzidine dye class).
Report on carcinogens : carcinogen profiles, 11, III106-III107 (2004-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service