Skip to Content
Merck
All Photos(1)

Key Documents

T2695

Supelco

Tetrandrine

analytical standard, for drug analysis

Synonym(s):

(1β)-6,6′,7,12-Tetramethoxy-2,2′-dimethylberbaman

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H42N2O6
CAS Number:
Molecular Weight:
622.75
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

Assay

≥90%

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

COc1ccc2C[C@@H]3N(C)CCc4cc(OC)c(OC)c(Oc5cc6[C@H](Cc7ccc(Oc1c2)cc7)N(C)CCc6cc5OC)c34

InChI

1S/C38H42N2O6/c1-39-15-13-25-20-32(42-4)34-22-28(25)29(39)17-23-7-10-27(11-8-23)45-33-19-24(9-12-31(33)41-3)18-30-36-26(14-16-40(30)2)21-35(43-5)37(44-6)38(36)46-34/h7-12,19-22,29-30H,13-18H2,1-6H3/t29-,30-/m0/s1

InChI key

WVTKBKWTSCPRNU-KYJUHHDHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrandrine (TET) is classified under the bisbenzylisoquinoline alkaloid family of compounds extracted from Stephania tetrandra S. Moore.

Application

Analgesic, antipyretic.
TET may be used as an analytical reference standard for the determination of TET in:
  • Blood samples by online multidimensional solid phase extraction liquid chromatography coupled to tandem mass spectrometry (MD-SPE-LC-MS/MS) equipped with electrospray ionization (ESI) source and multiple reaction monitoring (MRM) detection.
  • Rat plasma by ultra-high performance liquid chromatography (UHPLC) combined with MS/MS with MRM detection.
  • Qi-Fang-Xi-Bi-Granules used in Chinese medicine by reversed phase HPLC equipped with ultraviolet (UV) detector.
  • Fangji Huangqi Tang Chinese medicine by UHPLC coupled to triple quadrupole mass spectrometry (QqQ-MS) with MRM detection.

It may be used as an internal standard for the determination of hernandezine in:
  • Rat plasma and tissues by UHPLC-MS/MS with MRM detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Development and validation of an on-line multidimensional SPE-LC-MS/MS method for the quantitation of Tetrandrine in blood samples.
Caglar S, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 988, 25-32 (2015)
C Y Kwan et al.
Life sciences, 68(7), 841-847 (2001-02-24)
The effects of tetrandrine (TET), a Ca2+ antagonist of bis-benzylisoquinoline alkaloid origin, on cultured single bovine pulmonary artery endothelial cells were examined using fluorescence ratio imaging and whole-cell attached patch-clamp techniques. Thapsigargin (TSG, 100 nM), a selective endoplasmic reticulum Ca2+-ATPase
Development and validation of an UHPLC-QqQ-MS technique for simultaneous determination of ten bioactive components in Fangji Huangqi Tang.
Wang X, et al.
Journal of Analytical Methods in Chemistry, 2016(8), 1328-1332 (2016)
Determination of a novel anticancer AMPK activator hernandezine in rat plasma and tissues with a validated UHPLC-MS/MS method: Application to pharmacokinetics and tissue distribution study.
Song Y, et al.
Journal of Pharmaceutical and Biomedical Analysis, 141(8), 132-139 (2017)
Simultaneous determination of fangchinoline and tetrandrine in Qi-Fang-Xi-Bi-granules by RP-HPLC.
Lu XG, et al.
Journal of Chromatographic Science, 53(8), 1328-1332 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service