Skip to Content
Merck
All Photos(1)

Documents

T2117

Millipore

Thiol broth

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

application(s)

clinical testing
food and beverages

microbiology

suitability

Haemophilus spp.
nonselective for Neisseria spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for Vibrio spp.

Application

Used for culturing microorganisms from body fluids containing penicillin, streptomycin or sulphonamides.

Components

Ingredients (g/L):
Proteose peptone, 10.00
Yeast extract, 5.00
Dextrose, 1.00
Sodium chloride, 5.00
Thiol compound, 8.00
p-Amino benzoic acid, 0.05

Preparation Note

Suspend 29g of Thiol Broth in 1000 ml of distilled water. Heat to boiling to dissolve the medium completely. Dispense in tubes or erlenmeyer flasks to a depth of 6 cm for neutralization of Penicillin or in shallow layers for neutralization of Streptomycin. Sterilize by autoclaving at 15 lbs. pressure (121 °C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service