Skip to Content
Merck
All Photos(3)

Key Documents

STS0210

Sigma-Aldrich

ECO BRIJ® C10

greener alternative

Synonym(s):

Brij® C10, Polyethylene glycol hexadecyl ether, Polyoxyethylene (10) cetyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H33(OCH2CH2)nOH, n~10
CAS Number:
Beilstein:
6450627
UNSPSC Code:
12165104
NACRES:
NA.28

description

non-ionic

Quality Level

form

solid

mol wt

average Mn ~683

greener alternative product characteristics

Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤3% water

mp

32-34 °C (lit.)

acid number

≤1.0 mg KOH/g

hydroxyl value

75‑90 mg KOH/g

density

0.977 g/mL at 25 °C (lit.)

HLB

12.9

greener alternative category

SMILES string

CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C56H114O21/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17-58-19-21-60-23-25-62-27-29-64-31-33-66-35-37-68-39-41-70-43-45-72-47-49-74-51-53-76-55-56-77-54-52-75-50-48-73-46-44-71-42-40-69-38-36-67-34-32-65-30-28-63-26-24-61-22-20-59-18-16-57/h57H,2-56H2,1H3

InChI key

NLMKTBGFQGKQEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ECO Brij® C10 is an ethoxylated natural fatty alcohol, a nonionic bio-based surfactant produced from naturally occurring straight chain cetyl alcohol. This naturally derived detergent offers a number of functional benefits including detergency, wetting and dispersing making it suitable for a range of applications in biochemical and biological research.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biobased surfactant and is aligned with the 7th principle of Green Chemistry "Use of Renewable Feedstocks" and the 10th principle "Design for Degradation".

Application

Brij® C10 is a surfactant that can be used for applications such as: improving the wetting of PEDOT:PSS which can further be used in the fabrication of thin film transistors, formation of reverse micelles for the synthesis of high yielding nanostructures, in the preparation of platinum electrode for fuel cell applications

Features and Benefits

  • 100 % Renewable
  • 100 % Bio-based
  • Certified to the USDA BioPreferred Program
  • Lower carbon footprint than petrochemical-based versions
  • High-purity chemical suitable for a wide variety of research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Brij is a registered trademark of Croda International PLC
ECO BRIJ is a registered trademark of Croda Inc.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strategy for preparation of transparent organic thin film transistors with PEDOT:PSS electrodes and a polymeric gate dielectric
Albrecht G, et al.
Materials Science in Semiconductor Processing, 40, 772-776 (2015)
Scalable synthesis of sub-100 nm hollow carbon nanospheres for energy storage applications
Zhao H, et al.
Nano Research, 11(4), 1822-1833 (2018)
Electrodeposited Mesoporous Pt for Direct Ethanol Fuel Cells Anodes
Doan N
International Journal of Electrochemical Science, 11(4), 7631-7643 (2016)
G Tataru et al.
International journal of pharmaceutics, 404(1-2), 83-93 (2010-11-16)
Magnetic micro- and nanoparticles based on ferrofluid (maghemite) were elaborated by inverse emulsion crosslinking of sodium salt of carboxymethylcellulose (CMCNa) and gelatin. Crosslinking was carried out with glutaric aldehyde within aqueous droplets dispersed into toluene in presence of surfactants. The
Xue Li et al.
Chemosphere, 78(6), 752-759 (2009-12-17)
Currently, soil contamination by polybrominated diphenyl ethers (PBDEs) at e-waste recycling sites in China is of significant environmental concern and requires the identification of appropriate remediation technologies. In this paper, photodestruction of a model PBDE congener (BDE-99) in selected UV/surfactant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service