SMB01377
2AB-labeled Dextran Ladder
Synonym(s):
2-Aminobenzamide dextran ladder standard, Glycan Ladder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.79
Recommended Products
Assay
>90% (HPLC)
Quality Level
form
powder or crystals
technique(s)
HPLC: suitable
mass spectrometry (MS): suitable
impurities
≤10% Unlabeled Dextran ladder
shipped in
2-8°C
storage temp.
−20°C
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service