Skip to Content
Merck
All Photos(1)

Key Documents

S1400000

Streptomycin sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Streptomycin sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H39N7O12 · 1.5 H2SO4
CAS Number:
Molecular Weight:
728.69
Beilstein:
3894995
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

streptomycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.CN[C@H]1[C@H](O)[C@@H](O)[C@H](CO)O[C@H]1O[C@H]2[C@@H](O[C@@H](C)[C@]2(O)C=O)O[C@@H]3[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]3NC(N)=N.CN[C@H]4[C@H](O)[C@@H](O)[C@H](CO)O[C@H]4O[C@H]5[C@@H](O[C@@H](C)[C@]5(O)C=O)O[C@@H]6[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]6NC(N)=N

InChI

1S/2C21H39N7O12.3H2O4S/c2*1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35;3*1-5(2,3)4/h2*4-18,26,29,31-36H,3H2,1-2H3,(H4,22,23,27)(H4,24,25,28);3*(H2,1,2,3,4)/t2*5-,6-,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,21+;;;/m00.../s1

InChI key

QTENRWWVYAAPBI-YZTFXSNBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Streptomycin sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Mode of Action: Streptomycin acts by inhibiting prokaryote protein synthesis by binding to the S12 protein of the 30S ribosomal subunit, thus preventing the transition from imitation complex to chain-elongating ribosome. This causes miscoding or inhibits initiation.

Mode of Resistance: A mutation in rpsL, a gene for S12 ribosomal protein, prevents binding of streptomycin to the ribosome. An aminoglycoside phosphotransferase also inactivates streptomycin.

Antimicrobial spectrum: Streptomycin acts against gram-negative and gram-positive bacteria.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Zheng et al.
Clinical & experimental metastasis, 31(8), 921-933 (2014-09-17)
In prostate cancer metastases to bone, cancer cell-derived cytokines stimulate RANKL expression by cells of the osteoblast lineage, which in turn activates osteoclastic bone resorption. However, it is unclear whether cells of the osteoblast lineage signal back to prostate cancer
Annalisa Milano et al.
Journal of the American College of Cardiology, 64(8), 745-756 (2014-08-26)
Familial forms of primary sinus bradycardia have sometimes been attributed to mutations in HCN4, SCN5A, and ANK2. In these studies, no structural cardiac alterations were reported in mutation carriers. However, a cluster of reports in the literature describe patients presenting
Xue Tan et al.
Experimental eye research, 131, 77-83 (2015-01-13)
As a part of the visual cycle, all-trans-retinol (all-trans-ROL), the major form of vitamin A in circulating blood, is transported to the retinal pigment epithelium (RPE). All-trans-ROL is essential for normal retina function. However, recent researches have shown that excessive
Hannes Uchtenhagen et al.
Journal of immunology (Baltimore, Md. : 1950), 192(12), 5802-5812 (2014-05-16)
Our knowledge of the binding sites for neutralizing Abs (NAb) that recognize a broad range of HIV-1 strains (bNAb) has substantially increased in recent years. However, gaps remain in our understanding of how to focus B cell responses to vulnerable
Zhigang Liu et al.
Neuro-oncology, 16(6), 787-799 (2014-01-29)
Supratentorial primitive neuroectodermal tumor (sPNET) is a malignant brain tumor with poor prognosis. New model systems that replicate sPNET's molecular subtype(s) and maintain cancer stem cell (CSC) pool are needed. A fresh surgical specimen of a pediatric sPNET was directly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service