Skip to Content
Merck
All Photos(1)

Documents

S0765000

Diclofenac sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Diclofenac sodium salt, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Cl2NNaO2
CAS Number:
Molecular Weight:
318.13
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diclofenac

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[O-]C(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl

InChI

1S/C14H11Cl2NO2.Na/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1

InChI key

KPHWPUGNDIVLNH-UHFFFAOYSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diclofenac sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Standard NSAID and cyclooxygenase (COX) inhibitor. Major metabolites are 4´-hydroxydiclofenac and 5´-hydroxydiclofenac. Has been used as substrate selective for CYP2C9.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shahinaze A Fouad et al.
International journal of pharmaceutics, 453(2), 569-578 (2013-06-26)
Microemulsion (ME) and poloxamer microemulsion-based gel (PMBG) were developed and optimized to enhance transport of diclofenac epolamine (DE) into the skin forming in-skin drug depot for sustained transdermal delivery of drug. D-optimal mixture experimental design was applied to optimize ME
Rachel P L van Swelm et al.
Toxicology and applied pharmacology, 269(2), 141-149 (2013-03-20)
Diclofenac (DF) is a widely used non-steroidal anti-inflammatory drug for the treatment of rheumatic disorders, but is often associated with liver injury. We applied urinary proteomic profiling using MALDI-TOF MS to identify biomarkers for DF-induced hepatotoxicity in mice. Female CH3/HeOUJIco
R E Small
Clinical pharmacy, 8(8), 545-558 (1989-08-01)
The pharmacology, pharmacokinetics, clinical efficacy, adverse effects, and dosage of diclofenac sodium are reviewed. Diclofenac, the first nonsteroidal anti-inflammatory agent (NSAID) to be approved that is a phenylacetic acid derivative, competes with arachidonic acid for binding to cyclo-oxygenase, resulting in
R N Brogden et al.
Drugs, 20(1), 24-48 (1980-07-01)
Diclofenac sodium, a phenylacetic acid derivative, is a non-steroidal, anti-inflammatory, analgesic agent advocated for use in rheumatoid arthritis, degenerative joint disease, ankylosing spondylitis and allied conditions, and in the treatment of pain resulting from minor surgery, trauma and dysmenorrhoea. Published
L K Dodgen et al.
Environmental pollution (Barking, Essex : 1987), 182, 150-156 (2013-08-06)
Many pharmaceutical and personal care products (PPCPs) and endocrine-disrupting chemicals (EDCs) are present in reclaimed water, leading to concerns of human health risks from the consumption of food crops irrigated with reclaimed water. This study evaluated the potential for plant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service