QC1488
Trace Metals 1 - WS
certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
CofA
current certificate can be downloaded
packaging
ampule of 20 mL
concentration
in 5% HNO3
technique(s)
AAS: suitable
ICP: suitable
application(s)
environmental
format
matrix material
storage temp.
2-30°C
Related Categories
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Packaging
Product is packaged as a 20 mL concentrate and contains the following analytes:
- Arsenic
- Beryllium
- Cadmium
- Chromium
- Copper
- Iron
- Lead
- Lithium
- Manganese
- Mercury
- Nickel
- Selenium
- Zinc
- Aluminum
Other Notes
Values of analytes vary lot to lot. Some lots may not contain all analytes.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service