Recommended Products
grade
certified reference material
CofA
current certificate can be downloaded
packaging
ampule of 20 mL
manufacturer/tradename
RTC QCI-034-3
concentration
100% in H2O
technique(s)
AAS: suitable
ICP: suitable
application(s)
environmental
format
matrix material
storage temp.
2-30°C
Related Categories
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Other Notes
Values of analytes vary lot to lot. Some lots may not contain all analytes.
Analyte
Description
Chromium(VI) oxide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service