Skip to Content
Merck
All Photos(1)

Key Documents

PHR2011

Supelco

Gabapentin Related Compound B

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(1-Cyanocyclohexyl)acetic acid, (1-CYANOCYCLOHEXYL)ACETIC ACID, 1-Cyanocyclohexaneacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO2
CAS Number:
Molecular Weight:
167.21
Beilstein:
11150802
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001348
traceable to USP 1287347

API family

gabapentin

form

powder

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

OC(=O)CC1(CCCCC1)C#N

InChI

1S/C9H13NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-6H2,(H,11,12)

InChI key

YGFXLCAKCKPSQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB4759 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiwoon Ra et al.
Water research, 184, 116123-116123 (2020-07-23)
The occurrence and fate of the popular pharmaceutical gabapentin (GBP) in the urban water cycle were investigated with a focus on its transformation during water chlorination. GBP was detected in all samples with average concentrations of 1285 ng/L (n = 24) for wastewater

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service