Skip to Content
Merck
All Photos(1)

Documents

PHR1796

Supelco

Imipenem

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Imipenem monohydrate, (5R,6S)-6-[(1R)-1-Hydroxyethyl]-3-[[2-[(iminomethyl)amino]ethyl]thio]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid monohydrate, Primaxin monohydrate, Tienam monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N3O4S · H2O
CAS Number:
Molecular Weight:
317.36
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. I0090000
traceable to USP 1337809

API family

imipenem

form

solid

CofA

current certificate can be downloaded

packaging

pkg of 200 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

storage temp.

-10 to -25°C

SMILES string

O.C[C@@H](O)[C@@H]1[C@H]2CC(SCCNC=N)=C(N2C1=O)C(O)=O

InChI

1S/C12H17N3O4S.H2O/c1-6(16)9-7-4-8(20-3-2-14-5-13)10(12(18)19)15(7)11(9)17;/h5-7,9,16H,2-4H2,1H3,(H2,13,14)(H,18,19);1H2/t6-,7-,9-;/m1./s1

InChI key

GSOSVVULSKVSLQ-JJVRHELESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Imipenem belongs to the class of carbapenems, a beta-lactam antibiotic, effective against aerobic and anaerobic bacteria. Carbapenems act by inhibiting the peptidoglycan synthesis involved in bacterial cell wall formation, thus causing the death of the target microorganism.

Application

Imipenem may be used as a pharmaceutical reference standard for the quantification of the analyte in biological samples and pharmaceutical formulations by various analytical techniques.

Biochem/physiol Actions

Imipenem monohydrate is a broad spectrum B-lactam antibiotic. It is a member of the carbapenem class of "magic bullet" antibiotics for severe infections.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0262 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipika Singhal et al.
Microbial drug resistance (Larchmont, N.Y.), 24(8), 1082-1088 (2018-02-07)
With the increasing threat of multidrug-resistant organisms, such as Acinetobacter baumannii, the polymyxin class of drugs (colistin and polymyxin B) has become popular in clinical practice. A better understanding of antimicrobial susceptibility testing methods for colistin and polymyxin B is
Zhen-Liang Wen et al.
Chinese medical journal, 132(10), 1179-1187 (2019-05-30)
Antibiotics are frequently used to treat critically ill patients, and its use is often accompanied by intestinal dysbiosis that might further lead to bacterial translocation (BT). Nevertheless, studies on the relationship between antibiotic therapy and BT are rare. In the
Determination of imipenem in human plasma, urine and tissue by high-performance liquid chromatography
Carlucci G, et al.
Journal of Pharmaceutical and Biomedical Analysis, 8(3), 283- 286 (1990)
Determination of imipenem in plasma by high-performance liquid chromatography for pharmacokinetic studies in patients
Garcia-Capdevila L, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 692(1), 127- 132 (1997)
Stability-indicating HPLC method development and validation for simultane-ous determination of cilastatin and imipenem in pharmaceutical dosage forms
Kishore MS and Rambabu C
International Journal of Research in Pharmaceutical Sciences, 8(4), 560-567 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service