Skip to Content
Merck
All Photos(1)

Key Documents

PHR1771

Supelco

Digoxin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Digoxin, 12β-Hydroxydigitoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H64O14
CAS Number:
Molecular Weight:
780.94
Beilstein:
77011
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. D1900000
traceable to USP 1200000

API family

digoxin

CofA

current certificate can be downloaded

packaging

pkg of 300 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

C[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]2[C@@H](O)C[C@@H](O[C@@H]2C)O[C@H]3[C@@H](O)C[C@@H](O[C@@H]3C)O[C@H]4CC[C@@]5(C)[C@H](CC[C@@H]6[C@@H]5C[C@@H](O)[C@]7(C)[C@H](CC[C@]67O)C8=CC(=O)OC8)C4

InChI

1S/C41H64O14/c1-19-36(47)28(42)15-34(50-19)54-38-21(3)52-35(17-30(38)44)55-37-20(2)51-33(16-29(37)43)53-24-8-10-39(4)23(13-24)6-7-26-27(39)14-31(45)40(5)25(9-11-41(26,40)48)22-12-32(46)49-18-22/h12,19-21,23-31,33-38,42-45,47-48H,6-11,13-18H2,1-5H3/t19-,20-,21-,23-,24+,25-,26-,27+,28+,29+,30+,31-,33+,34+,35+,36-,37-,38-,39+,40+,41+/m1/s1

InChI key

LTMHDMANZUZIPE-PUGKRICDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Digoxin is one of the digitalis glycosides, extensively utilized for the treatment of congestive heart failure (CHF). It exhibits positive inotropic effects, antiarrhythmic as well as vasoconstrictor actions. It is reported that digoxin and other cardiac glycosides could play important role in the prevention and/or treatment of proliferative diseases like cancer.[1]

Application

Digoxin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations[2][3][4] and plasma samples[1] by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

A cardiac glycoside, substrate for Pgp, upregulates Pgp expression and down regulates SXR (Steroid Xenobiotic Receptor).
A cardiac glycoside, substrate for Pgp.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8515 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic analysis of digoxin formulations.
Desta B and McErlane KM
Journal of Pharmaceutical Sciences, 71(7), 777-780 (1982)
Selective and quantitative analysis of 4-hydroxybenzoate preservatives by microemulsion electrokinetic chromatography.
Mahuzier PE, et al.
Journal of Chromatography A, 924(1-2), 465-470 (2001)
Simultaneous determination of digoxin and digitoxin by micellar electrokinetic chromatography and application to drug formulations.
Tseng HH, et al.
Journal of Separation Science, 26(18), 1693-1698 (2003)
A new, high-throughput high-performance liquid chromatographic/mass spectrometric assay for therapeutic level monitoring of digoxin in human plasma.
Vlase L, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 92(5), 1390-1395 (2009)
Angela R Buckalew et al.
Archives of toxicology, 94(3), 873-885 (2020-02-18)
The Fischer rat thyroid follicular cell line (FRTL-5) endogenously expresses the sodium-iodide symporter (NIS) and has been used to identify environmental chemicals that perturb thyroid hormone homeostasis by disruption of NIS-mediated iodide uptake. Previously, a high-throughput radioactive iodide uptake (RAIU)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service