Skip to Content
Merck
All Photos(1)

Key Documents

PHR1611

Supelco

Mercaptopurine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

6-Mercaptopurine monohydrate, 6-Purinethiol, 6-Thiohypoxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4S·H2O
CAS Number:
Molecular Weight:
170.19
Beilstein:
4012091
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 773
traceable to USP 1392002

API family

mercaptopurine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.S=C1NC=Nc2nc[nH]c12

InChI

1S/C5H4N4S.H2O/c10-5-3-4(7-1-6-3)8-2-9-5;/h1-2H,(H2,6,7,8,9,10);1H2

InChI key

WFFQYWAAEWLHJC-UHFFFAOYSA-N

Gene Information

human ... PPAT(5471)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Mercaptopurine is an anticancer and an immunosuppressive drug employed for the management of acute lymphocytic leukemia (ALL), chronic myeloid leukemia (CML), Crohn′s disease, etc. Its mode of action involves interference in the nucleic acid synthesis by the inhibition of purine metabolism.

Application

Mercaptopurine may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations, suspensions, and double drug liposomes by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3476 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

6-mercaptopurine and daunorubicin double drug liposomes?preparation, drug-drug interaction and characterization
Agrawal V, et al.
Journal of liposome research, 15(3-4), 141-155 (2005)
Effect of buffer and antioxidant on stability of a mercaptopurine suspension
Aliabadi HM, et al.
American Journal of Health-System Pharmacy, 65(5), 441-447 (2008)
DEVELOPMENT AND VALIDATION OF STABILITY INDICATING RP-HPLC METHOD FOR DETERMINATION OF MERCAPTOPURINE-AN ANTI CANCER DRUG IN PHARMACEUTICAL FORMULATIONS
Suman P, et al.
European Journal of Biomedical AND Pharmaceutical sciences, 5(5), 752-759 (2018)
Mepivacaine hydrochloride
USP42-NF37
United States Pharmacopeia/National Formulary, 40(5), 2750-2750 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service