Skip to Content
Merck
All Photos(1)

Documents

PHR1521

Supelco

Ceftiofur Sodium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Sodium Ceftiofur

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1098165

API family

ceftiofur

CofA

current certificate can be downloaded

packaging

ampule of 1 g

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

General description

Ceftiofur Sodium is a caphalosporin antibiotic, with activity similar to other beta-lactam antibiotics. Its mode of action involves the inhibition of the synthesis of bacterial cell wall, thereby leading to cell death.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3328 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Environmental fate of ceftiofur sodium, a cephalosporin antibiotic. Role of animal excreta in its decomposition.
Gilbertson J T, et al.
Journal of Agricultural and Food Chemistry, 38(3), 890-894 (1990)
Ceftiofur sodium, a broad-spectrum cephalosporin: evaluation in vitro and in vivo in mice.
JRY, et al.
American Journal of Veterinary Research, 48(7), 1050-1053 (1987)
Comparison of plasma pharmacokinetics and bioequivalence of ceftiofur sodium in cattle after a single intramuscular or subcutaneous injection.
Brown A S, et al.
Journal of Veterinary Pharmacology and Therapeutics, 23(5), 273-280 (2000)
Saunders Handbook of Veterinary Drugs - E-Book: Small and Large Animal (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service