Skip to Content
Merck
All Photos(1)

Documents

PHR1314

Supelco

p-Xylene

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1,4-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
1901563
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to NIST 3005
traceable to USP 1601849

vapor density

3.7 (vs air)

vapor pressure

9 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

984 °F

expl. lim.

7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.495 (lit.)

bp

138 °C (lit.)

mp

12-13 °C (lit.)

density

0.861 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cc1ccc(C)cc1

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3

InChI key

URLKBWYHVLBVBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

p-Xylene may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB8973 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-speed analysis of residual solvents by flow-modulation gas chromatography
Wittrig RE, et al.
Journal of Chromatography A, 1027(1-2), 75-82 (2004)
Semi-automated peak trapping recycle chromatography instrument for peak purity investigations
Trone MD, et al.
Journal of Chromatography A, 1133(1-2), 104-111 (2006)
Sergio Grunder et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(49), 15632-15649 (2012-10-24)
Molecular gauge blocks, based on 1-7, 9-11 paraxylene rings, have been synthesized as part of a homologous series of oligoparaxylenes (OPXs) with a view to providing a molecular tool box for the construction of nano architectures-such as spheres, cages, capsules
G Gallastegui et al.
Bioresource technology, 102(17), 7657-7665 (2011-07-01)
Interactions of toluene and p-xylene in air treatment biofilters packed with an inert filter media were studied. The effect of the inlet load of toluene, p-xylene and mixtures of both compounds on the biodegradation rate was analyzed in three lab-scale
Martin Kuss-Petermann et al.
Journal of the American Chemical Society, 134(30), 12844-12854 (2012-07-20)
A homologous series of four molecules in which a phenol unit is linked covalently to a rhenium(I) tricarbonyl diimine photooxidant via a variable number of p-xylene spacers (n = 0-3) was synthesized and investigated. The species with a single p-xylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service