Skip to Content
Merck
All Photos(1)

Documents

PHR1274

Supelco

Melamine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2,4,6-Triamino-1,3,5-triazine, sym-Triaminotriazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N6
CAS Number:
Molecular Weight:
126.12
Beilstein:
124341
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1379183

API family

melamine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Nc1nc(N)nc(N)n1

InChI

1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)

InChI key

JDSHMPZPIAZGSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Melamine is a nitrogen-rich chemical employed in the production of plastics, resins, laminates, adhesives as well as flame retardants. It may also be utilized as a fertilizer. It is one of the metabolites of cyromazine, which is approved for use as an insecticide on a wide range of vegetable crops.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3464 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Urinary tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

572.0 °F - closed cup

Flash Point(C)

300 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method development and validation for melamine and its derivatives in rice concentrates by liquid chromatography. Application to animal feed samples
Mu?iz-Valencia R, et al.
Analytical and Bioanalytical Chemistry, 392(3), 523-531 (2008)
Hui Yang et al.
Nano letters, 15(3), 1730-1735 (2015-02-11)
We detect by optical microscopy Au and fluorescent nanoparticles (NPs) during their motion in water-based medium, using an array of dielectric microspheres that are patterned in a microwell array template. The microspheres act as lenses focusing the light originating from
Ana Paula Craig et al.
Annual review of food science and technology, 4, 369-380 (2013-01-10)
Surface-enhanced Raman spectroscopy (SERS) is an advanced Raman technique that enhances the vibrational spectrum of molecules adsorbed on or in the vicinity of metal particles and/or surfaces. Because of its readiness, sensitivity, and minimum sample preparation requirements, SERS is being
Lingjie Kong et al.
Optics letters, 38(2), 145-147 (2013-03-05)
Stimulated Raman scattering (SRS) microscopy allows label-free chemical imaging based on vibrational spectroscopy. Narrowband excitation with picosecond lasers creates the highest signal levels and enables imaging speeds up to video-rate, but it sacrifices chemical specificity in samples with overlapping bands
Yanhua Zhao et al.
Talanta, 113, 7-13 (2013-05-28)
Silver nanoparticles coated amino modified polystyrene microspheres (PS-NH2/Ag NPs) with extremely high surface enhanced Raman scattering (SERS) activity and uniform surface morphology were created by precise controlling of deposition time. Nanojets that were formed underneath the individual microspheres could be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service