Skip to Content
Merck
All Photos(1)

Key Documents

PHR1233

Supelco

Biotin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
Beilstein:
86838
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. B1116000
traceable to USP 1071508

API family

biotin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

231-233 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Biotin is a water-soluble vitamin, essential for amino acids and carbohydrates metabolism. It is involved in de novo synthesis of purine nucleotides and plays a role in gene expression and DNA replication.

Application

Biotin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical preparations using a bio-sensing system and chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0265 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Using of HPLC coupled with coulometric detector for the determination of biotin in pharmaceuticals.
Zerzanova A, et al.
Journal of Pharmaceutical and Biomedical Analysis, 45(5), 730-735 (2007)
Determination of biotin in foodstuffs and pharmaceutical preparations using a biosensing system based on the streptavidin--biotin interaction.
Reyes FD, et al.
Analytica Chimica Acta, 436(1), 109-117 (2001)
Liquid chromatographic determination of biotin in multivitamin-multimineral tablets.
Ekpe AE and Hazen C
Journal of Pharmaceutical and Biomedical Analysis, 16(8), 1311-1315 (1998)
Yuxi Feng et al.
PloS one, 6(2), e16865-e16865 (2011-03-08)
Vasoregression is a hallmark of vascular eye diseases but the mechanisms involved are still largely unknown. We have recently characterized a rat ciliopathy model which develops primary photoreceptor degeneration and secondary vasoregression. To improve the understanding of secondary vasoregression in
Sung Jae Shin et al.
Clinical and vaccine immunology : CVI, 15(8), 1277-1281 (2008-06-14)
We previously reported that protein antigens of serodiagnostic potential were more abundant in culture filtrates than cellular extracts from liquid cultures of Mycobacterium avium subsp. paratuberculosis (D. Cho and M. T. Collins, Clin. Vaccine Immunol. 13:1155-1161, 2006). Based on this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service