NIST928
Lead nitrate
NIST® SRM® 928
Synonym(s):
Lead(II) nitrate, Lead dinitrate
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
solid
packaging
pkg of 30 g
manufacturer/tradename
NIST®
mp
470 °C (dec.) (lit.)
application(s)
general analytical
format
neat
SMILES string
[PbH2++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
InChI
1S/2NO3.Pb/c2*2-1(3)4;/q2*-1;+2
InChI key
RLJMLMKIBZAXJO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
SRM 928_cert
SRM 928_SDS
Application
Other Notes
Lead Nitrate [Pb(NO3)2]
See certificate for values and more details at nist.gov/SRM.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 1A - Skin Sens. 1B - STOT RE 1
Target Organs
Blood,Central nervous system,Immune system,Kidney
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service