Skip to Content
Merck
All Photos(1)

Key Documents

NIST8040A

Sodium Oxalate Reductometric Standard

NIST®SRM®

Synonym(s):

Sodium oxalate, Ethanedioic acid sodium salt, Oxalic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCOONa
CAS Number:
Molecular Weight:
134.00
Beilstein:
3631622
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 60 g

manufacturer/tradename

NIST®

application(s)

general analytical

format

neat

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2

InChI key

ZNCPFRVNHGOPAG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

A unit of RM 8040a consists of one bottle containing 60 g of crystalline sodium oxalate. For more information, please refer to the COA and SDS.
SRM 8040A_cert

SRM 8040A_SDS

Application

The Reference Material (RM) consists of uniformly consistent, high-purity material intended for use as a working standard in oxidation-reduction reactions.

Features and Benefits

  • The non-certified values for the reductometric assay are expressed as mass fractions and are metrologically traceable to the materials and methods used in its determination.
  • Maintenance and expiration of RM value assignments, instructions for storage drying and use are provided on NIST certificate.

Other Notes

Reference/Informational value is provided for the following analytes.
Sodium Oxalate (Na2C2O4)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service