Skip to Content
Merck
All Photos(1)

Key Documents

NIST1810A

Linerboard

NIST® SRM® 1810a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 50 ea

manufacturer/tradename

NIST®

application(s)

general analytical

format

matrix material

General description

This Standard Reference Material (SRM) is intended to provide a uniform source of linerboard for use with ASTM® D2860 Standard Test Method for Adhesion of Pressure-Sensitive Tape to a Foberboard at 90 Degrees Angle and Constant Stress. For more information, please refer to the COA and SDS.

SRM 1810A_cert

SRM 1810A_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service