NIST114R
Portland Cement
NIST® SRM® 114r, fineness standard
Synonym(s):
Portland cement
Select a Size
Select a Size
About This Item
Recommended Products
grade
certified reference material
Agency
NIST
packaging
pkg of 20 vials (each containing 5g of cement)
manufacturer/tradename
NIST®
format
matrix material
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
SRM 114R_cert
SRM 114R_SDS
Application
Analysis Note
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service