Skip to Content
Merck
All Photos(1)

Key Documents

N2515

Supelco

D-Norpropoxyphene maleate salt

analytical standard

Synonym(s):

1,2-Diphenyl-3-methyl-4-(methylamino)-2-butyl propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27NO2 · C4H4O4
CAS Number:
Molecular Weight:
441.52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)\C=C/C(O)=O.CCC(=O)OC(Cc1ccccc1)(C(C)CNC)c2ccccc2

InChI

1S/C21H27NO2.C4H4O4/c1-4-20(23)24-21(17(2)16-22-3,19-13-9-6-10-14-19)15-18-11-7-5-8-12-18;5-3(6)1-2-4(7)8/h5-14,17,22H,4,15-16H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

HCQPFYNZJNOOKN-BTJKTKAUSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Lang-Jensen et al.
Pharmacology & toxicology, 64(2), 228-232 (1989-02-01)
In a double blind cross-over study 10 healthy male volunteers were given either 300 mg dextropropoxyphene napsylat (DP) or placebo daily for 16 days. The serum levels of DP and the metabolite nordextropropoxyphene were measured on day 3, 6 and
R S Wu et al.
Bioconjugate chemistry, 8(3), 385-390 (1997-05-01)
The synthesis of [S-(R,S)]-4-[[methyl[2-methyl-3-(1-oxopropoxy)-3, 4-diphenylbutyl]amino]-1-oxobutoxy]-2,5-pyrrolidinedione+ ++ (propoxyphene active ester, 2) is described. This was used as an intermediate to prepare a propoxyphene immunogen, [S-(R,S)]-4-[methyl][2-methyl-3-(1-oxopropoxy)-3,4-diphenylbuty l]-amino]- 1-oxobutyl-Bovine Thyroglobulin (3). This immunogen was then used to generate antibodies which demonstrate good cross-reactivity
P Kintz et al.
Journal de toxicologie clinique et experimentale, 10(2), 89-94 (1990-03-01)
A method for the identification and quantification of dextropropoxyphene, norpropoxyphene, and methaqualone in plasma by GC/NPD is presented. The procedure employs cyclizine as the internal standard and requires no derivatization. After a single-step extraction, analysis is achieved in 8 min.
V Rigourd et al.
Journal of analytical toxicology, 32(9), 787-789 (2008-11-22)
Dextropropoxyphene (DP) and norpropoxyphene (NP) are commonly used in the treatment of postpartum pain. The drug is widely prescribed in Europe and Canada and has been recently approved for use in the U.S. Its safety during breastfeeding, however, has not
F T Horrigan et al.
The Journal of general physiology, 107(2), 243-260 (1996-02-01)
Voltage-dependent ionic currents were recorded from squid giant fiber lobe neurons using the whole-cell patch-clamp technique. When applied to the bathing solution, methadone was found to block IK, I Na and I Ca. Both I Na and I Ca were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service