Skip to Content
Merck
All Photos(1)

Documents

N0950000

Nitrofural

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Nitro-2-furaldehyde semicarbazone, 5-Nitro-2-furfural semicarbazone, Furacin, NFZ, Nitrofurazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O4
CAS Number:
Molecular Weight:
198.14
Beilstein:
86403
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nitrofural

manufacturer/tradename

EDQM

mp

242-244 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)N\N=C\c1ccc(o1)[N+]([O-])=O

InChI

1S/C6H6N4O4/c7-6(11)9-8-3-4-1-2-5(14-4)10(12)13/h1-3H,(H3,7,9,11)/b8-3+

InChI key

IAIWVQXQOWNYOU-FPYGCLRLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nitrofural EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Muta. 2 - Repr. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

male reproductive organs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan He et al.
Analytica chimica acta, 1141, 1-12 (2020-11-30)
In the last few years, sildenafil and its analogues have emerged as an illegal ingredient in health food and products, causing illness and death. To address this problem, herein, a gold nanoparticle-based immunochromatographic assay for simultaneous detection sildenafil and its
Jian-Xin Huang et al.
Biomolecules, 9(12) (2019-11-28)
In this work, a monoclonal antibody-based indirect competitive enzyme-linked immunosorbent assay (icELISA) was established to detect tylosin and tilmicosin in milk and water samples. A sensitive and specific monoclonal antibody was prepared by rational designed hapten, which was achieved by
Characterization of the genotoxic properties of nitrofurans: nitrofurazone and furazolidone.
M Kobierska-Szeliga et al.
Acta biochimica Polonica, 41(1), 1-5 (1994-01-01)
[Therapeutically used 5-nitrofural derivatives. Review of literature. 2].
H Kala et al.
Die Pharmazie, 26(4), 193-207 (1971-04-01)
Nitrofural (nitrofurazone).
IARC monographs on the evaluation of carcinogenic risks to humans, 50, 195-209 (1990-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service