Skip to Content
Merck
All Photos(1)

Key Documents

IRMM644

Sulfur-32S solution

IRMM®, certified reference material, Spike Isotopic Reference Material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
32S
CAS Number:
Molecular Weight:
31.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

isotopic purity

 32S

manufacturer/tradename

JRC

concentration

326.28 μmol/kg±0.80 μmol/kg 32S in 3.2 M HNO3

application(s)

general analytical

format

single component solution

mass shift

depleted

SMILES string

[32S]

InChI

1S/S/i1+0

InChI key

NINIDFKCEFEMDL-IGMARMGPSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
IRMM644

Legal Information

IRMM is a registered trademark of European Commission

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beata Olas et al.
Chemico-biological interactions, 220, 20-24 (2014-06-15)
Hydrogen sulfide (H2S) is a well known toxic gas at high levels. However, at physiological levels, H2S may play a role in the pathogenesis of various cardiovascular diseases. The objective was to study the effects of exogenous H2S on the
Dae-Hwan Kim et al.
Nanoscale, 6(23), 14549-14554 (2014-10-29)
A high-quality Sb₂S₃ thin-absorber with controllable thickness was reproducibly formed by atomic layer deposition (ALD) technique. Compared with conventional chemical bath deposition (CBD), the Sb₂S₃ absorber deposited by ALD did not contain oxide or oxygen impurities and showed a very
Jean-Roch Mouret et al.
Journal of agricultural and food chemistry, 63(12), 3271-3278 (2015-03-13)
The knowledge of gas-liquid partitioning of aroma compounds during winemaking fermentation could allow optimization of fermentation management, maximizing concentrations of positive markers of aroma and minimizing formation of molecules, such as hydrogen sulfide (H2S), responsible for defects. In this study
Jean-Michel Paquette et al.
Clinical therapeutics, 36(11), 1650-1664 (2014-09-17)
Trimebutine 3-thiocarbamoylbenzenesulfonate (GIC-1001) is a new drug intended to be used for the management of visceral pain in patients undergoing sedation-free, full colonoscopy. The objectives of this Phase I, single-center, randomized, double-blinded, placebo-controlled, integrated study were to evaluate the safety
Xu Zheng et al.
PloS one, 10(3), e0119780-e0119780 (2015-03-18)
In response to interleukin 6 (IL-6) stimulation, both CD45RO and CD45RB, but not CD45RA, translocate to lipid rafts. However, the significance of this distinct translocation and the downstream signals in CD45 isoforms-participated IL-6 signal are not well understood. Using sucrose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service