Skip to Content
Merck
All Photos(1)

Key Documents

I0320800

Iohexol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-[N-(2,3-Dihydroxypropyl)acetamido]-2,4,6-triiodo-N,N′-bis(2,3-dihydroxypropyl)isophthalamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26I3N3O9
CAS Number:
Molecular Weight:
821.14
Beilstein:
2406632
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

iohexol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

IC1=C(C(NCC(O)CO)=O)C(I)=C(C(NCC(CO)O)=O)C(I)=C1N(CC(CO)O)C(C)=O

InChI

1S/C19H26I3N3O9/c1-8(29)25(4-11(32)7-28)17-15(21)12(18(33)23-2-9(30)5-26)14(20)13(16(17)22)19(34)24-3-10(31)6-27/h9-11,26-28,30-32H,2-7H2,1H3,(H,23,33)(H,24,34)

InChI key

NTHXOOBQLCIOLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iohexol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Van de Kelft et al.
Neurosurgery, 28(4), 570-574 (1991-04-01)
Intracranial hemorrhage is an uncommon complication of dural puncture. In most instances, hematomas are subdural; they may be unilateral or bilateral. Rarely are intraparenchymal cerebral hemorrhages related to dural puncture. This report describes a delayed occurrence of bilateral intraparenchymal hemorrhages
Catrina Hansen et al.
AJR. American journal of roentgenology, 202(4), W357-W364 (2014-03-26)
The purpose of this study was to evaluate the diagnostic performance of a dedicated dual-source dual-energy MDCT (DECT) protocol for the detection of urothelial tumors. A DECT protocol including furosemide and split-bolus contrast injection was used in 69 consecutive patients
N J Bureau et al.
AJNR. American journal of neuroradiology, 35(8), 1467-1474 (2014-05-31)
Transforaminal corticosteroid injections can be performed in the management of cervical radiculopathy but carry the risk of catastrophic complications. This study compares the efficacy of transforaminal and facet corticosteroid injections at 4 weeks' follow-up. We randomly assigned 56 subjects to
Jay Smith et al.
Journal of ultrasound in medicine : official journal of the American Institute of Ultrasound in Medicine, 34(1), 83-93 (2014-12-30)
To determine the feasibility and accuracy of sonographically guided posterior subtalar joint (PSTJ) injections performed through the sinus tarsi. A single experienced operator completed 10 sonographically guided PSTJ injections via the sinus tarsi on 10 unembalmed cadaveric ankle-foot specimens. Injections
Robert P Jones et al.
AJR. American journal of roentgenology, 204(3), 519-526 (2015-02-26)
OBJECTIVE. The purpose of this article is to evaluate the hypothesis that Alvarado scores of 3 or lower identify adult patients who are unlikely to benefit from CT after appendiceal ultrasound fails to show the appendix and is otherwise normal.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service