Skip to Content
Merck
All Photos(1)

Key Documents

G0550000

Gramicidin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

gramicidin

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[nH]1c2c(c(c1)C[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](NC(=O)[C@@H](NC(=O)[C@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC=O)C(C)C)C)CC(C)C)C)C(C)C)C(C)C)C(C)C)C(=O)N[C@H](CC(C)C)C(=O)N[C@@H](Cc7c8c([nH]c7)cccc8)C(=O)N[C@H](CC(C)C)C(=O)N[C@@H](Cc5c6c([nH]c5)

InChI

1S/C99H140N20O17/c1-51(2)37-73(109-86(123)59(17)107-81(122)49-105-96(133)82(55(9)10)106-50-121)89(126)108-60(18)87(124)117-84(57(13)14)98(135)119-85(58(15)16)99(136)118-83(56(11)12)97(134)116-80(44-64-48-104-72-34-26-22-30-68(64)72)95(132)112-76(40-54(7)8)92(129)115-79(43-63-47-103-71-33-25-21-29-67(63)71)94(131)111-75(39-53(5)6)91(128)114-78(42-62-46-102-70-32-24-20-28-66(62)70)93(130)110-74(38-52(3)4)90(127)113-77(88(125)100-35-36-120)41-61-45-101-69-31-23-19-27-65(61)69/h19-34,45-48,50-60,73-80,82-85,101-104,120H,35-44,49H2,1-18H3,(H,100,125)(H,105,133)(H,106,121)(H,107,122)(H,108,126)(H,109,123)(H,110,130)(H,111,131)(H,112,132)(H,113,127)(H,114,128)(H,115,129)(H,116,134)(H,117,124)(H,118,136)(H,119,135)/t59-,60-,73+,74+,75+,76+,77-,78-,79-,80-,82-,83-,84+,85-/m0/s1

InChI key

ZWCXYZRRTRDGQE-SORVKSEFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O S Andersen et al.
Indian journal of biochemistry & biophysics, 33(5), 331-342 (1996-10-01)
The linear gramicidins are peptide antibiotics that form cation-selective channels in lipid bilayers. Gramicidin channels have very well-defined functional characteristics, and the structure of membrane-spanning gramicidin A channels is known at atomic resolution. These features make the gramicidins well suited
[Gramicidin S: characteristics of a producer, cultivation conditions, and interactions with a producer].
N S Egorov et al.
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic], 48(12), 24-28 (2004-06-05)
[Structural and functional characteristics of gramicidin S in connection with its antibiotic activity].
A N Polin et al.
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic], 48(12), 29-32 (2004-06-05)
N Akaike
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 113(6), 339-347 (1999-10-19)
Cl- is one of the major ionic constituents of cells and extracellular spaces. Intracellular Cl- plays an important role in regulating the cell volume and pH, in both salt secretion and reabsorption, in membrane excitability, and G-protein-dependent intracellular signal transduction.
Benoît Roux
Accounts of chemical research, 35(6), 366-375 (2002-06-19)
Ion channels are highly specific membrane-spanning protein structures which serve to facilitate the passage of selected ions across the lipid barrier. In the past decade, molecular dynamics simulations based on atomic models and realistic microscopic interactions with explicit solvent and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service