Skip to Content
Merck
All Photos(1)

Key Documents

G0525000

Gonadorelin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H75N17O13
CAS Number:
Molecular Weight:
1182.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

gonadorelin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[nH]1cnc(c1)CC(NC(=O)C6NC(=O)CC6)C(=O)NC(Cc4c5c([nH]c4)cccc5)C(=O)NC(CO)C(=O)NC(Cc3ccc(cc3)O)C(=O)NCC(=O)NC(CC(C)C)C(=O)NC(CCCNC(=N)N)C(=O)N2C(CCC2)C(=O)NCC(=O)N.OC(=O)C.O

InChI

1S/C55H75N17O13.C2H4O2.H2O/c1-29(2)19-38(49(80)67-37(9-5-17-60-55(57)58)54(85)72-18-6-10-43(72)53(84)62-25-44(56)75)66-46(77)26-63-47(78)39(20-30-11-13-33(74)14-12-30)68-52(83)42(27-73)71-50(81)40(21-31-23-61-35-8-4-3-7-34(31)35)69-51(82)41(22-32-24-59-28-64-32)70-48(79)36-15-16-45(76)65-36;1-2(3)4;/h3-4,7-8,11-14,23-24,28-29,36-43,61,73-74H,5-6,9-10,15-22,25-27H2,1-2H3,(H2,56,75)(H,59,64)(H,62,84)(H,63,78)(H,65,76)(H,66,77)(H,67,80)(H,68,83)(H,69,82)(H,70,79)(H,71,81)(H4,57,58,60);1H3,(H,3,4);1H2

InChI key

OOBWZCWPKJOGPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Gonadorelin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung-Hoon Kim et al.
The Journal of reproductive medicine, 59(1-2), 63-68 (2014-03-07)
To evaluate the effects of combined administration of gonadotropin-releasing hormone (GnRH) agonist with human chorionic gonadotropin (hCG)for final oocyte maturation in GnRH antagonist cycles for in vitro fertilization (IVF). A total of 120 infertile women undergoing GnRH antagonist multiple-dose protocol
Sihyun Cho et al.
Acta obstetricia et gynecologica Scandinavica, 93(1), 38-44 (2014-03-08)
Although the levonorgestrel-releasing intrauterine system (LNG-IUS) is effective in reducing the recurrence of endometriosis-associated pain, its efficacy in preventing endometrioma recurrence is questionable. We compared the efficacy of postoperative use of LNG-IUS with oral contraceptives (OC) for preventing endometrioma recurrence.
Jack M Zuckerman et al.
Urology, 83(3), 670-674 (2013-12-24)
To evaluate for a possible testosterone surge during transition of therapy from degarelix to leuprolide. We conducted an investigator-initiated, prospective, single-arm, open-label trial for evaluation of a potential testosterone surge during a transition of therapy from degarelix to leuprolide. Study
Rebecca M Perrett et al.
The Journal of biological chemistry, 289(11), 7873-7883 (2014-02-01)
Gonadotropin-releasing hormone (GnRH) is secreted in brief pulses that stimulate synthesis and secretion of pituitary gonadotropin hormones and thereby mediate control of reproduction. It acts via G-protein-coupled receptors to stimulate effectors, including ERK. Information could be encoded in GnRH pulse
Senji Hoshi et al.
Gan to kagaku ryoho. Cancer & chemotherapy, 41(1), 65-69 (2014-01-16)
The efficacy and safety of degarelix, a luteinizing hormone-releasing hormone(LH-RH)antagonist, in patients with prostate cancer(PCa)were evaluated in a phase II, open-label, multicenter clinical trial. In this trial, a total of 13 patients were accrued at the Yamagata Prefectural Central Hospital

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service