Skip to Content
Merck
All Photos(1)

Documents

ERMBC211

Rice flour (Total As and As species)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

food and beverages

format

matrix material

storage temp.

−20°C

General description

Certified for the mass fractions of total arsenic, the sum of arsenite/arsenate and dimethylarsinic acid

Analysis Note

For more information please see:
ERMBC211

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Trace Elements / Metallorganics: Dimethylarsinic acid, Total Arsenic (As), The sum of arsenite and arsenate

Matrix Group: Nuts, Soy, Edible Oils and Fats

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arsenic in rice and other foods--reply.
Ana Navas-Acien et al.
JAMA internal medicine, 174(2), 298-299 (2014-02-05)
A Sommella et al.
Environmental pollution (Barking, Essex : 1987), 181, 38-43 (2013-07-03)
Rice is comparatively efficient at assimilating inorganic arsenic (Asi), a class-one, non-threshold carcinogen, into its grain, being the dominant source of this element to mankind. Here it was investigated how the total arsenic (Ast) and Asi content of Italian rice
Sandra Munera-Picazo et al.
Journal of food science, 79(1), T122-T128 (2013-12-10)
Celiac disease is an autoimmune disease that affects the villi of the small intestine causing abdominal pain, gas, diarrhea, or bad absorption due to gluten intolerance. The only treatment for this disease consists of a lifelong gluten free diet; this
Fang-Jie Zhao et al.
Environmental science & technology, 47(9), 3957-3966 (2013-03-26)
Rice is a major source of inorganic arsenic (iAs) in the human diet because paddy rice is efficient at accumulating As. Rice As speciation is dominated by iAs and dimethylarsinic acid (DMA). Here we review the global pattern in rice
Stanislav Musil et al.
Analytical chemistry, 86(2), 993-999 (2013-12-21)
Because of the toxicity of inorganic arsenic (iAs), only iAs needs to be monitored in food and feedstuff. This demands the development of easy and quick analytical methods to screen large number of samples. This work focuses on hydride generation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service