Skip to Content
Merck
All Photos(2)

Key Documents

E3768

Sigma-Aldrich

(−)-Epigallocatechin

≥95% (HPLC), from green tea

Synonym(s):

(−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxyflavane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O7
CAS Number:
Molecular Weight:
306.27
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.21

biological source

green tea

Quality Level

Assay

≥95% (HPLC)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3cc(O)c(O)c(O)c3

InChI

1S/C15H14O7/c16-7-3-9(17)8-5-12(20)15(22-13(8)4-7)6-1-10(18)14(21)11(19)2-6/h1-4,12,15-21H,5H2/t12-,15-/m1/s1

InChI key

XMOCLSLCDHWDHP-IUODEOHRSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Epigallocatechin is one of the main catechin compounds found in green tea and the most abundant one (~20%), with antioxidant properties.

Application

(−)-Epigallocatechin (EGC) has been used:
  • as a reference standard in high-performance liquid chromatography (HPLC) analysis
  • to analyze its inhibitory effect on α-amylase and α-glucosidase activity to control postprandial hyperglycemia in Diabetes mellitus
  • in cell viability assays to check its efficacy and protection against ferroptosis and iron toxicity in murine pancreatic β-cells
  • to evaluate its potential anti-inflammatory effects on long-term acute and chronic Pristen-induced arthritis
  • to treat pregnant and nursing mice, on in vitro angiogenesis, proliferation, and movement of endothelial cells
  • to evaluate its effect on pregnant and nursing mice spleen based on the number and localization of CD4+ (cluster of differentiation 4) and CD8+ cluster of differentiation 8) cells

Biochem/physiol Actions

(−)-Epigallocatechin (EGC) has proven to be an anti-cancer agent by its inhibitory effects on heregulin-b1 (HRG)-induced migration/invasion in MCF-7 human breast carcinoma cells. It has also shown strong antiviral activity against the herpes simplex virus.
Green tea polyphenol. Anticancer agent. Lower antioxidant activity than EGCG.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukihiro Kushima et al.
Biological & pharmaceutical bulletin, 32(5), 899-904 (2009-05-08)
Migration/invasion is involved in the multiple steps of metastasis, resulting in a poor prognosis of breast cancer. (-)-Epigallocatechin-3-gallate (EGCG) in green tea inhibits the metastasis of some cancer cell lines. Cell migration/invasion assays using Boyden chambers demonstrated that (-)-epigallocatechin (EGC)
Sławomir Lewicki et al.
Central-European journal of immunology, 42(1), 10-16 (2017-07-07)
Rhodiola kirilowii, a member of Crassulaceae family, grows wildly in Asiatic mountains and is also cultivated in some European countries. Its underground parts traditionally are used for enhance physical and mental performance of the body. In our previous papers we
Jun Xu et al.
Molecules (Basel, Switzerland), 22(8) (2017-08-15)
Over the centuries, infectious diseases caused by viruses have seriously threatened human health globally. Viruses are responsible not only for acute infections but also many chronic infectious diseases. To prevent diseases caused by viruses, the discovery of effective antiviral drugs
Evaluation of alpha-amylase and alpha-glucosidase inhibitory activity of flavonoids
Gu C, et al.,
International Journal of Food and Nutrition Science, 2(6), 1-6 (2015)
Robert Zdanowski et al.
Central-European journal of immunology, 42(1), 17-23 (2017-07-07)
Angiogenesis plays an important role in many physiological processes, among them the formation of tissues and organs during embryogenesis. A lot of medicinal plants exhibit angiomodulatory properties. This creates the need for a thorough check of whether the plant extracts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service