Skip to Content
Merck
All Photos(1)

Key Documents

E2150000

Ethosuximide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Ethyl-2-methylsuccinimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO2
CAS Number:
Molecular Weight:
141.17
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ethosuximide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1NC(CC1(C)CC)=O

InChI

1S/C7H11NO2/c1-3-7(2)4-5(9)8-6(7)10/h3-4H2,1-2H3,(H,8,9,10)

InChI key

HAPOVYFOVVWLRS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethosuximide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E B Posner et al.
The Cochrane database of systematic reviews, (3)(3), CD003032-CD003032 (2003-08-15)
Absence seizures are brief epileptic seizures which present in childhood and adolescence. They are characterised by sudden loss of awareness and an electroencephalogram (EEG) typically shows generalised spike wave discharges at three cycles per second. Ethosuximide, valproate and lamotrigine are
E B Posner et al.
The Cochrane database of systematic reviews, (4)(4), CD003032-CD003032 (2005-10-20)
Absence seizures are brief epileptic seizures which present in childhood and adolescence. They are characterised by sudden loss of awareness and an electroencephalogram (EEG) typically shows generalised spike wave discharges at three cycles per second. Ethosuximide, valproate and lamotrigine are
G V Massey et al.
Pediatric neurology, 11(1), 59-61 (1994-07-01)
We describe a case of aplastic anemia in an 8-year-old girl which was diagnosed 8 months after initiation of ethosuximide as treatment for absence seizures. Blood counts had been previously monitored and were normal. The patient successfully underwent allogeneic bone
S J Wallace
Neurologic clinics, 4(3), 601-616 (1986-08-01)
Ethosuximide is the drug of first choice in absence seizures. Idiosyncratic side effects are rare. Valproate is effective against absence, generalized tonic-colonic, and partial seizures. It is the drug of choice in photosensitive seizures. Valproate is, on the whole, free
M Zafer Gören et al.
CNS drug reviews, 13(2), 224-239 (2007-07-14)
Ethosuximide, 2-ethyl-2-methylsuccinimide, has been used extensively for "petit mal" seizures and it is a valuable agent in studies of absence epilepsy. In the treatment of epilepsy, ethosuximide has a narrow therapeutic profile. It is the drug of choice in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service