Skip to Content
Merck
All Photos(5)

Documents

E1769

Sigma-Aldrich

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride

BioXtra

Synonym(s):

N-Ethyl-N′-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDAC, EDC, EDC hydrochloride, WSC hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17N3 · HCl
CAS Number:
Molecular Weight:
191.70
Beilstein:
5764110
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product name

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra

product line

BioXtra

Quality Level

form

powder

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

mp

110-115 °C (lit.)

solubility

H2O: 0.5 M, clear to very slightly hazy, colorless to very faintly yellow

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

−20°C

SMILES string

Cl.CCN=C=NCCCN(C)C

InChI

1S/C8H17N3.ClH/c1-4-9-8-10-6-5-7-11(2)3;/h4-7H2,1-3H3;1H

InChI key

FPQQSJJWHUJYPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride has been used:
  • for post-fixation and crosslinking of small ribonucleic acid (RNAs) species from formalin fixed paraffin embedded (FFPE) tissues, for fluorescent in situ hybridisation (FISH) and immunofluorescence (IF) signals
  • for cross-linking of nanofiber matrices with mouse lung extract
  • for activation of substrate XNA oligonucleotides

Biochem/physiol Actions

Water soluble condensing reagent. EDAC is generally utilized as a carboxyl activating agent for amide bonding with primary amines. In addition, it will react with phosphate groups. EDAC has been used in peptide synthesis; crosslinking proteins to nucleic acids; and preparation of immunoconjugates as examples. Typically, EDAC is utilized in the pH range 4.0-6.0 without buffers. In particular, amine and carboxylate buffers should be avoided.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Stomach,large intestine,lymph node

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organ-derived coatings on electrospun nanofibers as ex vivo microenvironments
Fischer SN, et al.
Biomaterials, 32(2), 538-546 (2011)
Directed evolution of artificial enzymes (XNAzymes) from diverse repertoires of synthetic genetic polymers
Taylor AI and Holliger P
Nature Protocols, 10(10), 1625-1625 (2015)
Henry H Lee et al.
Nature communications, 10(1), 2383-2383 (2019-06-05)
DNA is an emerging medium for digital data and its adoption can be accelerated by synthesis processes specialized for storage applications. Here, we describe a de novo enzymatic synthesis strategy designed for data storage which harnesses the template-independent polymerase terminal
Amrita D Chaudhuri et al.
Frontiers in cellular neuroscience, 7, 160-160 (2013-09-26)
Identification of the cell type of origin for normal or aberrant gene expression is critical for many studies, and poses a significant problem for some regulatory RNAs such as microRNAs. MicroRNAs are small non-coding RNAs that regulate cellular function by
Francesca Paradiso et al.
Frontiers in bioengineering and biotechnology, 7, 343-343 (2020-01-11)
A fundamental structural component of extracellular matrix in all connective and interstitial tissue, collagen is the most abundant protein in the human body. To date, mammalian collagens sources represent the golden standard for multiple biomedical applications, while marine-derived collagens have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service