Skip to Content
Merck
All Photos(1)

Key Documents

E1620000

Estradiol hemihydrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

β-Estradiol hemihydrate, 1,3,5-Estratriene-3,17β-diol, 3,17β-Dihydroxy-1,3,5(10)-estratriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O2 · 0.5H2O
CAS Number:
Molecular Weight:
281.39
Beilstein:
1914275
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

estradiol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2O.C[C@]56CC[C@H]7[C@@H](CCc8cc(O)ccc78)[C@@H]5CC[C@@H]6O

InChI

1S/2C18H24O2.H2O/c2*1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20;/h2*3,5,10,14-17,19-20H,2,4,6-9H2,1H3;1H2/t2*14-,15-,16+,17+,18+;/m11./s1

InChI key

ZVVGLAMWAQMPDR-WVEWYJOQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Estradiol hemihydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Piet et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(26), 10828-10839 (2013-06-28)
Circulating gonadal steroid hormones are thought to modulate a wide range of brain functions. However, the effects of steroid fluctuations through the ovarian cycle on the intrinsic properties of neurons are not well understood. We examined here whether gonadal steroids
Yuko Hara et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(1), 486-491 (2013-12-04)
Humans and nonhuman primates are vulnerable to age- and menopause-related decline in working memory, a cognitive function reliant on the energy-demanding recurrent excitation of neurons within Brodmann's Area 46 of the dorsolateral prefrontal cortex (dlPFC). Here, we tested the hypothesis
Aniruddha Das et al.
Archives of sexual behavior, 43(7), 1403-1415 (2014-03-07)
Using nationally representative data from the 2005-2006 U.S. National Social Life, Health, and Aging Project, this study queried relationship, sexual, and sex hormone patterns among married evangelical women and men aged 57-85, relative to those in other religions. Results suggested
Shuzo Hirata et al.
Nature materials, 13(10), 938-946 (2014-09-10)
In materials showing reverse saturable absorption (RSA), the optical absorbance increases as the power of the light incident on them increases. To date, RSA has only been observed when very intense light sources, such as short-pulse lasers, are used. Here
Angelo Di Leo et al.
Journal of the National Cancer Institute, 106(1), djt337-djt337 (2013-12-10)
At the time of the initial analysis of overall survival (OS) for the Comparison of Faslodex in Recurrent or Metastatic Breast Cancer (CONFIRM) randomized, double-blind, phase III trial, approximately 50% of patients had died. A final analysis of OS was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service