Skip to Content
Merck
All Photos(1)

Key Documents

D5121

Sigma-Aldrich

Acculong DNA Polymerase

suitable for PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.54

form

liquid

Quality Level

technique(s)

PCR: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Acculong DNA Polymerase has improved abilities of long fragment amplification, amplification specificity, and amplification yield with the addition of unique elongation factors, specific promotion factors and plateau release inhibitors.
This DNA Polymerase proves to be greatly effective with both simple and complex templates.
For simple templates, it can amplify fragments up to 40 kb. For complex templates, it can amplify fragments up to 20 kb; using cDNA templates, it can effectively amplify fragments up to 10 kb.
Acculong DNA Polymerase also has good resistance to PCR inhibitors and can be used for direct PCR of bacteria, fungi, plant tissues, animal tissues or whole blood samples.

Features and Benefits

Acculong DNA Polymerase has the following advantages:




  • High Fidelity: 52 times that of Taq Polymerase and 6 times that of Pfu DNA polymerase.
  • Increased Speed: Amplification speed is 4-150 times more than conventional polymerases.
  • Optimized buffer system applicable to a wide range of templates.
  • Time Saver: Ready-to-use premix thereby reducing experimental steps.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service