Skip to Content
Merck
All Photos(1)

Key Documents

D2880000

Dirithromycin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

[9S(R)]-9-Deoxo-11-deoxy-9,11-[imino[2-(2-methoxy)ethylidene]oxy]erthromycin; LY-237216

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H78N2O14
CAS Number:
Molecular Weight:
835.07
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dirithromycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)C4N[C@@H](COCCOC)OC([C@H]4C)[C@]1(C)O

InChI

1S/C42H78N2O14/c1-15-29-42(10,49)37-24(4)32(43-30(56-37)21-52-17-16-50-13)22(2)19-40(8,48)36(58-39-33(45)28(44(11)12)18-23(3)53-39)25(5)34(26(6)38(47)55-29)57-31-20-41(9,51-14)35(46)27(7)54-31/h22-37,39,43,45-46,48-49H,15-21H2,1-14H3/t22-,23-,24+,25+,26-,27+,28+,29-,30-,31+,32+,33-,34+,35+,36-,37-,39+,40-,41+,42-/m1/s1

InChI key

WLOHNSSYAXHWNR-KZYCBHIHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dirithromycin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macrolide - induced clinically relevant drug interactions with cytochrome P-450A (CYP) 3A4: an update focused on clarithromycin, azithromycin and dirithromycin.
J F Westphal
British journal of clinical pharmacology, 50(4), 285-295 (2000-09-30)
R N Brogden et al.
Drugs, 48(4), 599-616 (1994-10-01)
Dirithromycin is a new macrolide with a spectrum and degree of in vitro antimicrobial activity similar to that of erythromycin. Compared with erythromycin, dirithromycin has a long elimination half-life enabling once-daily administration, and it also achieves a greater cellular:extracellular concentration
S M Wintermeyer et al.
The Annals of pharmacotherapy, 30(10), 1141-1149 (1996-10-01)
To review the clinical microbiology and therapeutic use of dirithromycin, emphasizing comparative data between dirithromycin and the standard macrolide erythromycin, as well as clarithromycin and azithromycin. A MEDLINE search of English-language literature during the years 1966-1996, and an extensive review
G D Sides et al.
The Journal of antimicrobial chemotherapy, 31 Suppl C, 65-75 (1993-03-01)
Dirithromycin is a new member of the macrolide class of antibiotics and has been developed for oral administration. Dirithromycin is a 14-membered lactone ring macrolide and is the C9-oxazine derivative of erythromycylamine. The human pharmacokinetics and clinical pharmacology of dirithromycin
V S Watkins et al.
The Annals of pharmacotherapy, 31(3), 349-356 (1997-03-01)
To describe the drug interactions of dirithromycin, a new macrolide, and to compare them with those of other macrolides. A literature search was performed using MEDLINE to identify articles published between January 1980 and July 1995 concerning the drug interactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service